Search
| Components | Molecular Weight | Concentration (mg/L) | mM |
|---|---|---|---|
| Amino Acids | |||
| L-Arginine hydrochloride | 211.0 | 1260.0 | 5.971564 |
| L-Cystine | 240.0 | 240.0 | 1.0 |
| L-Histidine hydrochloride-H2O | 210.0 | 420.0 | 2.0 |
| L-Isoleucine | 131.0 | 520.0 | 3.9694657 |
| L-Leucine | 131.0 | 520.0 | 3.9694657 |
| L-Lysine hydrochloride | 183.0 | 720.5 | 3.9371586 |
| L-Methionine | 149.0 | 150.0 | 1.0067114 |
| L-Phenylalanine | 165.0 | 320.0 | 1.939394 |
| L-Threonine | 119.0 | 480.0 | 4.0336137 |
| L-Tryptophan | 204.0 | 100.0 | 0.49019608 |
| L-Tyrosine | 181.0 | 360.0 | 1.9889503 |
| L-Valine | 117.0 | 460.0 | 3.931624 |
| Vitamins | |||
| Choline chloride | 140.0 | 10.0 | 0.071428575 |
| D-Calcium pantothenate | 477.0 | 10.0 | 0.02096436 |
| Folic Acid | 441.0 | 10.0 | 0.022675738 |
| Niacinamide | 122.0 | 10.0 | 0.08196721 |
| Pyridoxal hydrochloride | 204.0 | 10.0 | 0.04901961 |
| Riboflavin | 376.0 | 1.0 | 0.0026595744 |
| Thiamine hydrochloride | 337.0 | 10.0 | 0.029673591 |
| i-Inositol | 180.0 | 20.0 | 0.11111111 |
| Inorganic Salts | |||
| Calcium Chloride (CaCl2-2H2O) | 147.0 | 2640.0 | 17.959183 |
| Magnesium Sulfate (MgSO4-7H2O) | 246.0 | 2000.0 | 8.130081 |
| Potassium Chloride (KCl) | 75.0 | 4000.0 | 53.333332 |
| Sodium Chloride (NaCl) | 58.0 | 68000.0 | 1172.4138 |
| Sodium Phosphate monobasic (NaH2PO4-2H2O) | 156.0 | 1580.0 | 10.128205 |
| Other Components | |||
| D-Glucose (Dextrose) | 180.0 | 10000.0 | 55.555557 |
| Phenol Red | 376.4 | 100.0 | 0.26567483 |
11430 - MEM, 10x, no glutamine
Cat No:
11430030
10370 - MEM, NEAA, no glutamine
Cat No:
10370021
,
10370088
12360 - MEM, HEPES, no glutamine
Cat No:
12360038
51200 - MEM, no glutamine, no phenol red
Cat No:
51200038
,
51200046
32360 - MEM, HEPES, no glutamine
Cat No:
32360026
,
32360034
19993 - MEM Rega-3
Cat No:
19993013