Technical Resources
21430 - MEM, 10x, no glutamine
ComponentsMolecular WeightConcentration (mg/L)mM
Amino Acids
L-Arginine hydrochloride211.01260.05.971564
L-Cystine240.0240.01.0
L-Histidine hydrochloride-H2O210.0420.02.0
L-Isoleucine131.0520.03.9694657
L-Leucine131.0520.03.9694657
L-Lysine hydrochloride183.0720.53.9371586
L-Methionine149.0150.01.0067114
L-Phenylalanine165.0320.01.939394
L-Threonine119.0480.04.0336137
L-Tryptophan204.0100.00.49019608
L-Tyrosine181.0360.01.9889503
L-Valine117.0460.03.931624
Vitamins
Choline chloride140.010.00.071428575
D-Calcium pantothenate477.010.00.02096436
Folic Acid441.010.00.022675738
Niacinamide122.010.00.08196721
Pyridoxal hydrochloride204.010.00.04901961
Riboflavin376.01.00.0026595744
Thiamine hydrochloride337.010.00.029673591
i-Inositol180.020.00.11111111
Inorganic Salts
Calcium Chloride (CaCl2-2H2O)147.02640.017.959183
Magnesium Sulfate (MgSO4-7H2O)246.02000.08.130081
Potassium Chloride (KCl)75.04000.053.333332
Sodium Chloride (NaCl)58.068000.01172.4138
Sodium Phosphate monobasic (NaH2PO4-2H2O)156.01580.010.128205
Other Components
D-Glucose (Dextrose)180.010000.055.555557
Phenol Red376.4100.00.26567483
Reference:
Related Products:
11430 - MEM, 10x, no glutamine
Cat No: 11430030
10370 - MEM, NEAA, no glutamine
Cat No: 10370021 , 10370088
12360 - MEM, HEPES, no glutamine
Cat No: 12360038
51200 - MEM, no glutamine, no phenol red
Cat No: 51200038 , 51200046
32360 - MEM, HEPES, no glutamine
Cat No: 32360026 , 32360034
19993 - MEM Rega-3
Cat No: 19993013