Components | Molecular Weight | Concentration (mg/L) | mM |
---|---|---|---|
Amino Acids | |||
Glycine | 75.0 | 50.0 | 0.6666667 |
L-Alanine | 89.0 | 25.0 | 0.28089887 |
L-Arginine | 211.0 | 105.0 | 0.49763033 |
L-Asparagine-H2O | 150.0 | 25.0 | 0.16666667 |
L-Aspartic acid | 133.0 | 30.0 | 0.22556391 |
L-Cysteine hydrochloride-H2O | 176.0 | 100.0 | 0.5681818 |
L-Cystine | 313.0 | 31.0 | 0.09904154 |
L-Glutamic Acid | 147.0 | 75.0 | 0.5102041 |
L-Glutamine | 146.0 | 292.0 | 2.0 |
L-Histidine | 155.0 | 31.0 | 0.2 |
L-Isoleucine | 131.0 | 52.4 | 0.4 |
L-Leucine | 131.0 | 52.0 | 0.39694658 |
L-Lysine | 183.0 | 73.0 | 0.3989071 |
L-Methionine | 149.0 | 15.0 | 0.10067114 |
L-Phenylalanine | 165.0 | 32.0 | 0.19393939 |
L-Proline | 115.0 | 40.0 | 0.3478261 |
L-Serine | 105.0 | 25.0 | 0.23809524 |
L-Threonine | 119.0 | 48.0 | 0.40336135 |
L-Tryptophan | 204.0 | 10.0 | 0.04901961 |
L-Tyrosine | 181.0 | 36.0 | 0.19889502 |
L-Valine | 117.0 | 46.0 | 0.3931624 |
Vitamins | |||
Ascorbic Acid | 176.0 | 50.0 | 0.2840909 |
Biotin | 244.0 | 0.1 | 4.0983607E-4 |
Choline chloride | 140.0 | 1.0 | 0.007142857 |
D-Calcium pantothenate | 477.0 | 1.0 | 0.002096436 |
Folic Acid | 441.0 | 1.0 | 0.0022675737 |
Niacinamide | 122.0 | 1.0 | 0.008196721 |
Pyridoxal hydrochloride | 204.0 | 1.0 | 0.004901961 |
Riboflavin | 376.0 | 0.1 | 2.6595744E-4 |
Thiamine hydrochloride | 337.0 | 1.0 | 0.002967359 |
Vitamin B12 | 1355.0 | 1.36 | 0.0010036901 |
i-Inositol | 180.0 | 2.0 | 0.011111111 |
Inorganic Salts | |||
Calcium Chloride (CaCl2-2H2O) | 147.0 | 264.0 | 1.7959183 |
Magnesium Sulfate (MgSO4) (anhyd.) | 120.0 | 97.67 | 0.8139166 |
Potassium Chloride (KCl) | 75.0 | 400.0 | 5.3333335 |
Sodium Bicarbonate (NaHCO3) | 84.0 | 2200.0 | 26.190475 |
Sodium Chloride (NaCl) | 58.0 | 6800.0 | 117.24138 |
Sodium Phosphate monobasic (NaH2PO4-H2O) | 138.0 | 140.0 | 1.0144928 |
Ribonucleosides | |||
Adenosine | 267.0 | 10.0 | 0.037453182 |
Cytidine | 243.0 | 10.0 | 0.041152265 |
Guanosine | 283.0 | 10.0 | 0.03533569 |
Uridine | 244.0 | 10.0 | 0.040983606 |
Deoxyribonucleosides | |||
2'Deoxyadenosine | 251.0 | 10.0 | 0.03984064 |
2'Deoxycytidine HCl | 264.0 | 11.0 | 0.041666668 |
2'Deoxyguanosine | 267.0 | 10.0 | 0.037453182 |
Thymidine | 242.0 | 10.0 | 0.041322313 |
Other Components | |||
D-Glucose (Dextrose) | 180.0 | 1000.0 | 5.5555553 |
Lipoic Acid | 206.0 | 0.2 | 9.708738E-4 |
Phenol Red | 376.4 | 10.0 | 0.026567481 |
Sodium Pyruvate | 110.0 | 110.0 | 1.0 |
Reference:
Related Products:
12561 - MEM alpha, no nucleosides
Cat No:
12561049
,
12561056
,
12561072
12571 - MEM alpha, nucleosides
Cat No:
12571048
,
12571063
,
12571071
,
12571089
41061 - MEM alpha, nucleosides, no phenol red
Cat No:
41061029
,
41061037
22561, MEM alpha, no nucleosides
Cat No:
22561021
,
22561054
A10490 - MEM alpha, nucleosides, no ascorbic acid
Cat No:
A1049001