Technical Resources
Medium 106
A sterile, liquid medium for the culture of human dermal fibroblasts. This basal medium requires the addition of LSGS (S-003-10) or LSGS Kit (S-003-K) prior to use. Below is the composition list. The concentrations are proprietary.
ComponentsMolecular WeightConcentration (mg/L)mM
Amino Acids
Glycine75.0confidentialn/a
L-Alanine89.0confidentialn/a
L-Arginine hydrochloride211.0confidentialn/a
L-Asparagine-H2O150.0confidentialn/a
L-Aspartic acid133.0confidentialn/a
L-Cysteine121.0confidentialn/a
L-Glutamic Acid147.0confidentialn/a
L-Glutamine146.0confidentialn/a
L-Histidine hydrochloride-H2O210.0confidentialn/a
L-Isoleucine131.0confidentialn/a
L-Leucine131.0confidentialn/a
L-Lysine hydrochloride183.0confidentialn/a
L-Methionine149.0confidentialn/a
L-Phenylalanine165.0confidentialn/a
L-Proline115.0confidentialn/a
L-Serine105.0confidentialn/a
L-Threonine119.0confidentialn/a
L-Tryptophan204.0confidentialn/a
L-Tyrosine181.0confidentialn/a
L-Valine117.0confidentialn/a
Vitamins
Choline chloride140.0confidentialn/a
D-Pantothenic Acid238.0confidentialn/a
Folinic Acid512.0confidentialn/a
Myo-Inositol180.0confidentialn/a
Niacinamide122.0confidentialn/a
Pyridoxal hydrochloride206.0confidentialn/a
Riboflavin376.0confidentialn/a
Thiamine hydrochloride337.0confidentialn/a
Vitamin B121355.0confidentialn/a
d-Biotin244.0confidentialn/a
Inorganic Salts
Ammonium Metavanadate NaVO3117.0confidentialn/a
Ammonium Molybdate ((NH4)6Mo7O24-4H2O)1236.0confidentialn/a
Calcium Chloride (CaCl2-2H2O)147.0confidentialn/a
Cupric sulfate (CuSO4-5H2O)250.0confidentialn/a
Ferric sulfate (FeSO4-7H2O)278.0confidentialn/a
Magnesium Sulfate (MgSO4-7H2O)247.0confidentialn/a
Manganese Sulfate (MnSO4-H20)169.0confidentialn/a
Nickelous Chloride NiCl2 6H2O238.0confidentialn/a
Potassium Phosphate monobasic (KH2PO4)136.0confidentialn/a
Sodium Bicarbonate (NaHCO3)84.0confidentialn/a
Sodium Chloride (NaCl)58.0confidentialn/a
Sodium Meta Silicate Na2SiO3 9H2O284.0confidentialn/a
Sodium Selenite (Na2SeO3)173.0confidentialn/a
Tin Chloride (SnCl2.2H2O)226.0confidentialn/a
Zinc sulfate (ZnSO4-7H2O)288.0confidentialn/a
Other Components
Adenine.HCl172.0confidentialn/a
D-Glucose (Dextrose)180.0confidentialn/a
DL-alpha-Lipoic Acid206.0confidentialn/a
HEPES238.0confidentialn/a
Phenol Red376.4confidentialn/a
Putrescine 2HCl161.0confidentialn/a
Sodium Pyruvate110.0confidentialn/a
Thymidine242.0confidentialn/a
Reference:
Related Products:
Medium 131
Cat No: M131500
Medium 154
Cat No: M154500
Medium 154
Cat No: M154CF500
Medium 154
Cat No: M154CFPRF500
EpiLife® Medium
Cat No: MEPI500CA
EpiLife® Medium
Cat No: MEPICF500
EpiLife® Medium
Cat No: MEPICFPRF500
Medium 231
Cat No: M231500
Medium 200
Cat No: m200500
Medium 200
Cat No: M200PRF500
Medium 254
Cat No: M254500
Medium 254
Cat No: M254CF500
Medium 171
Cat No: M171500