Search
A sterile, concentrated (20X) solution intended for use with Medium 231 to culture human smooth muscle cells. Below is the composition list. The concentrations are proprietary.
Catalog Number(s)
S00725| Components | Molecular Weight | Concentration (mg/L) | mM |
|---|---|---|---|
| Proteins | |||
| BSA | confidential | n/a | |
| Basic FGF | confidential | n/a | |
| EGF | confidential | n/a | |
| Insulin | confidential | n/a | |
| Other Components | |||
| FBS | confidential | n/a | |
| Heparin | confidential | n/a | |
Human Keratinocyte Growth Supplement (HKGS)
Cat No:
s0015
Human Melanocyte Growth Supplement (HMGS)
Cat No:
s0025
Low Serum Growth Supplement (LSGS)
Cat No:
s00310
Microvascular Growth Supplement (MVGS)
Cat No:
s00525
Smooth Muscle Differentiation Supplement (SMDS)
Cat No:
s0085
Human Corneal Growth Supplement (HCGS)
Cat No:
s0095
EpiLife® Defined Growth Supplement (EDGS)
Cat No:
s0125
Mammary Epithelial Growth Supplement (MEGS)
Cat No:
s0155
S0165 - Human Melanocyte Growth Supplement-2, PMA-Free (HMGS-2)
Cat No:
s0165