Technical Resources
25050 - 0.25% Trypsin (1x), phenol red
ComponentsMolecular WeightConcentration (mg/L)mM
Inorganic Salts
Potassium Chloride (KCl)75.0400.05.3333335
Potassium Phosphate monobasic (KH2PO4)136.060.00.44117647
Sodium Bicarbonate (NaHCO3)84.0350.04.1666665
Sodium Chloride (NaCl)58.08000.0137.93103
Sodium Phosphate dibasic (Na2HPO4-7H2O)268.090.00.33582088
Other Components
D-Glucose (Dextrose)180.01000.05.5555553
Phenol Red398.010.00.025125628
Trypsin23800.02500.00.10504202
Reference:
Related Products:
25300 - 0.05% Trypsin - EDTA (1x), phenol red
Cat No: 25300054 , 25300062 , 25300096 , 25300104 , 25300120
25200 - 0.25% Trypsin-EDTA (1x), phenol red
Cat No: 25200056 , 25200072 , 25200114
15400 - 0.5% Trypsin-EDTA (10x), no phenol red
Cat No: 15400054
15050 - 0.25% Trypsin (1x), phenol red
Cat No: 15050057 , 15050065
15090 - 2.5% Trypsin (10x), no phenol red
Cat No: 15090046
TrypLE™ Select (1X), no phenol red
Cat No: 12563011 , 12563029
TrypLE™ Express (1X), no phenol red
Cat No: 12604013 , 12604021 , 12604039 , 12604054
TrypLE™ Select (10X), no phenol red
Cat No: A1217701 , A1217702 , A1217703
TrypLE™ Select CTS™
Cat No: A1285901
TrypLE™ Express (1X), phenol red
Cat No: 12605010 , 12605028 , 12605036 , 12605093