Search
| Components | Molecular Weight | Concentration (mg/L) | mM |
|---|---|---|---|
| Amino Acids | |||
| L-Arginine hydrochloride | 211.0 | 18.6 | 0.08815166 |
| L-Asparagine | 132.0 | 10.0 | 0.07575758 |
| L-Cystine 2HCl | 313.0 | 26.2 | 0.08370607 |
| L-Glutamine | 146.0 | 204.0 | 1.3972603 |
| L-Histidine hydrochloride-H2O | 210.0 | 66.0 | 0.31428573 |
| L-Isoleucine | 131.0 | 75.0 | 0.57251906 |
| L-Leucine | 131.0 | 30.0 | 0.22900763 |
| L-Lysine hydrochloride | 183.0 | 50.0 | 0.27322406 |
| L-Methionine | 149.0 | 100.0 | 0.67114097 |
| L-Phenylalanine | 165.0 | 60.0 | 0.36363637 |
| L-Serine | 105.0 | 15.0 | 0.14285715 |
| L-Threonine | 119.0 | 30.0 | 0.25210086 |
| L-Tryptophan | 204.0 | 10.0 | 0.04901961 |
| L-Tyrosine disodium salt dihydrate | 261.0 | 86.5 | 0.33141762 |
| L-Valine | 117.0 | 70.0 | 0.5982906 |
| L-Valine | 117.0 | 70.0 | 0.5982906 |
| Vitamins | |||
| Choline chloride | 140.0 | 1.5 | 0.010714286 |
| D-Calcium pantothenate | 477.0 | 0.5 | 0.001048218 |
| Folic Acid | 441.0 | 10.0 | 0.022675738 |
| Niacinamide | 122.0 | 0.5 | 0.0040983604 |
| Pyridoxal hydrochloride | 204.0 | 0.5 | 0.0024509805 |
| Riboflavin | 376.0 | 0.5 | 0.0013297872 |
| Thiamine hydrochloride | 337.0 | 1.0 | 0.002967359 |
| d-Biotin | 244.0 | 0.01 | 4.0983607E-5 |
| i-Inositol | 180.0 | 1.5 | 0.008333334 |
| Inorganic Salts | |||
| Calcium Chloride (CaCl2) (anhyd.) | 111.0 | 68.7 | 0.6189189 |
| Magnesium Chloride (anhydrous) | 95.0 | 47.0 | 0.49473685 |
| Potassium Chloride (KCl) | 75.0 | 400.0 | 5.3333335 |
| Sodium Bicarbonate (NaHCO3) | 84.0 | 1125.0 | 13.392858 |
| Sodium Chloride (NaCl) | 58.0 | 8000.0 | 137.93103 |
| Sodium Phosphate dibasic (Na2HPO4) anhydrous | 268.0 | 59.9 | 0.22350746 |
| Sodium Phosphate monobasic (NaH2PO4-H2O) | 138.0 | 69.0 | 0.5 |
| Other Components | |||
| D-Glucose (Dextrose) | 180.0 | 1000.0 | 5.5555553 |
| Phenol Red | 376.4 | 5.0 | 0.013283741 |