Search Thermo Fisher Scientific
Catalog Number(s)
Components | Molecular Weight | Concentration (mg/L) | mM |
---|---|---|---|
Amino Acids | |||
Glycine | 75.0 | 100.0 | 1.3333334 |
L-Alanine | 89.0 | 50.0 | 0.56179774 |
L-Arginine hydrochloride | 211.0 | 140.0 | 0.6635071 |
L-Aspartic acid | 133.0 | 60.0 | 0.45112783 |
L-Cysteine hydrochloride-H2O | 176.0 | 0.2 | 0.0011363636 |
L-Cystine 2HCl | 240.0 | 52.0 | 0.21666667 |
L-Glutamic Acid | 147.0 | 150.0 | 1.0204082 |
L-Glutamine | 146.0 | 200.0 | 1.369863 |
L-Histidine hydrochloride-H2O | 210.0 | 53.76 | 0.25599998 |
L-Hydroxyproline | 131.0 | 20.0 | 0.15267175 |
L-Isoleucine | 131.0 | 80.0 | 0.610687 |
L-Leucine | 131.0 | 120.0 | 0.9160305 |
L-Lysine hydrochloride | 183.0 | 140.0 | 0.76502734 |
L-Methionine | 149.0 | 30.0 | 0.20134228 |
L-Phenylalanine | 165.0 | 50.0 | 0.3030303 |
L-Proline | 115.0 | 80.0 | 0.6956522 |
L-Serine | 105.0 | 50.0 | 0.47619048 |
L-Threonine | 119.0 | 60.0 | 0.5042017 |
L-Tryptophan | 204.0 | 20.0 | 0.09803922 |
L-Tyrosine disodium salt dihydrate | 261.0 | 116.0 | 0.44444445 |
L-Valine | 117.0 | 50.0 | 0.42735043 |
Vitamins | |||
Ascorbic Acid | 176.0 | 0.1 | 5.681818E-4 |
Biotin | 244.0 | 0.02 | 8.196721E-5 |
Choline chloride | 140.0 | 1.0 | 0.007142857 |
D-Calcium pantothenate | 477.0 | 0.02 | 4.192872E-5 |
Folic Acid | 441.0 | 0.02 | 4.5351473E-5 |
Menadione (Vitamin K3) | 172.0 | 0.02 | 1.16279065E-4 |
Niacinamide | 122.0 | 0.05 | 4.0983607E-4 |
Nicotinic acid (Niacin) | 123.0 | 0.05 | 4.0650408E-4 |
Para-Aminobenzoic Acid | 137.0 | 0.1 | 7.2992704E-4 |
Pyridoxal hydrochloride | 204.0 | 0.05 | 2.4509805E-4 |
Pyridoxine hydrochloride | 206.0 | 0.05 | 2.4271845E-4 |
Riboflavin | 376.0 | 0.02 | 5.319149E-5 |
Thiamine hydrochloride | 337.0 | 0.02 | 5.934718E-5 |
Vitamin A (acetate) | 328.0 | 0.2 | 6.097561E-4 |
Vitamin D2 (Calciferol) | 397.0 | 0.2 | 5.0377834E-4 |
i-Inositol | 180.0 | 0.1 | 5.5555557E-4 |
Inorganic Salts | |||
Calcium Chloride (CaCl2-2H2O) | 147.0 | 400.0 | 2.7210884 |
Ferric nitrate (Fe(NO3)-9H2O) | 404.0 | 1.4 | 0.0034653465 |
Magnesium Sulfate (MgSO4-7H2O) | 246.0 | 195.34 | 0.794065 |
Potassium Chloride (KCl) | 75.0 | 800.0 | 10.666667 |
Sodium Bicarbonate (NaHCO3) | 84.0 | 2500.0 | 29.761906 |
Sodium Chloride (NaCl) | 58.0 | 13600.0 | 234.48276 |
Sodium Phosphate dibasic (Na2HPO4) anhydrous | 138.0 | 316.0 | 2.289855 |
Other Components | |||
2-deoxy-D-ribose | 134.0 | 1.0 | 0.0074626864 |
Adenine sulfate | 404.0 | 20.0 | 0.04950495 |
Adenosine 5'-phosphate | 347.0 | 0.4 | 0.0011527378 |
Adenosine 5'-triphosphate | 605.0 | 2.0 | 0.0033057851 |
Cholesterol | 387.0 | 0.4 | 0.0010335917 |
D-Glucose (Dextrose) | 180.0 | 2000.0 | 11.111111 |
Glutathione (reduced) | 307.0 | 0.1 | 3.257329E-4 |
Guanine hydrochloride | 188.0 | 0.6 | 0.0031914895 |
Hypoxanthine | 136.0 | 0.8 | 0.005882353 |
Ribose | 150.0 | 1.0 | 0.006666667 |
Sodium acetate-3H2O | 136.0 | 100.0 | 0.7352941 |
Thymine | 126.0 | 0.6 | 0.004761905 |
Tween 80® | 40.0 | Infinity | |
Uracil | 112.0 | 0.6 | 0.005357143 |
Xanthine | 174.0 | 0.68 | 0.003908046 |
1. Morgan, J.F. and Campbell, M.E. (1955) J. Natl. Cancer Inst., 16:557.
2. Morgan, J.F., Morton, H.J. and Parker R.C. (1950) Proc. Soc. Exp. Biol. Med., 73:1.
12340 - Medium 199, HEPES
Cat No:
12340030
11043 - Medium 199, no phenol red
Cat No:
11043023
11825 - Medium 199, 10x
Cat No:
11825015
22340 - Medium 199, HEPES
Cat No:
22340020
,
22340087
21180 - Medium 199, 10x
Cat No:
21180021
41150 - Medium 199, GlutaMAX(TM)
Cat No:
41150020
,
41150087
31153 - Medium 199, low sodium bicarbonate
Cat No:
31153026