Technical Resources - Media Formulations
R001100 - 0.025% Trypsin - EDTA Solution
  • Catalog Number(s)
ComponentsMolecular WeightConcentration (mg/L)mM
Inorganic Salts
Potassium Chloride (KCl)75.01900.025.333334
Potassium Phosphate monobasic (KH2PO4)136.01900.013.970589
Sodium Bicarbonate (NaHCO3)84.0350.04.1666665
Sodium Chloride (NaCl)58.07600.0131.03448
Sodium Phosphate dibasic (Na2HPO4-H2O)142.01092.07.6901407
Other Components
D-Glucose (Dextrose)180.01000.05.5555553
EDTA328.0100.00.30487806
Phenol Red398.01.20.0030150756
Trypsin23800.0250.00.010504202
Verweis
Related Products:
25300 - 0.05% Trypsin - EDTA (1x), phenol red
Cat No: 25300054 , 25300062 , 25300096 , 25300104 , 25300120
25200 - 0.25% Trypsin-EDTA (1x), phenol red
Cat No: 25200056 , 25200072 , 25200114
15400 - 0.5% Trypsin-EDTA (10x), no phenol red
Cat No: 15400054
15050 - 0.25% Trypsin (1x), phenol red
Cat No: 15050057 , 15050065
15090 - 2.5% Trypsin (10x), no phenol red
Cat No: 15090046
25050 - 0.25% Trypsin (1x), phenol red
Cat No: 25050014 , 25050030
TrypLE™ Select (1X), no phenol red
Cat No: 12563011 , 12563029
TrypLE™ Express (1X), no phenol red
Cat No: 12604013 , 12604021 , 12604039 , 12604054
TrypLE™ Select (10X), no phenol red
Cat No: A1217701 , A1217702 , A1217703
TrypLE™ Select CTS™
Cat No: A1285901
TrypLE™ Express (1X), phenol red
Cat No: 12605010 , 12605028 , 12605036 , 12605093