Search Thermo Fisher Scientific
Gibco® TAP Media is optimized for the growth and maintenance of Chlamydomonas reinhardtii. The formulation comes ready-to-use at 1X to let you avoid laborious media preparation steps.
Each Gibco® TAP Media comes in the award winning Gibco® bottle designed for easier use in the biosafety cabinet, minimizing the risk of contamination, and helping you perform cell culture more consistently. Together, the superior packaging and quality, greater reliability, and improved consistency in Chlamydomonas culture, results in better overall efficiency and more robust data
Components | Molecular Weight | Concentration (mg/L) | mM |
---|---|---|---|
Inorganic Salts | |||
Ammonium Chloride | 53.49 | 380.0 | 7.104131 |
Ammonium Molybdate ((NH4)6Mo7O24-4H2O) | 1235.86 | 10.0 | 0.008091532 |
Boric Acid (H3BO3) | 61.83 | 10.0 | 0.16173378 |
Calcium Chloride (CaCl2-2H2O) | 147.0 | 50.0 | 0.34013605 |
Cobalt Chloride (CoCl2-6H2O) | 237.93 | 20.0 | 0.08405834 |
Cupric sulfate (CuSO4-5H2O) | 249.69 | 20.0 | 0.08009932 |
Ferric sulfate (FeSO4-7H2O) | 278.01 | 50.0 | 0.17984964 |
Magnesium Sulfate (MgSO4-7H2O) | 246.47 | 100.0 | 0.40572888 |
Manganese chloride (MnCl2-4H2O) | 197.9 | 50.0 | 0.25265285 |
Potassium Phosphate monobasic (KH2PO4) | 136.09 | 50.0 | 0.36740392 |
Potassium phosphate dibasic (K2HPO4) | 110.0 | Infinity | |
Zinc sulfate (ZnSO4-7H2O) | 287.55 | 20.0 | 0.06955312 |
Other Components | |||
EDTA | 372.24 | 50.0 | 0.13432194 |
Glacial Acetic Acid | 60.05 | 1100.0 | 18.31807 |
Tris | 121.14 | 2420.0 | 19.976887 |
Terrific Broth
Cat No:
22711022
2-YT Broth
Cat No:
22712020
Gibco® BG-11 Media: Optimized for Cyanobacteria
Cat No:
a1379901
,
A1379902