Di-Methyl-Histone H3 (Lys79) Antibody (720079) in FACS
Flow Cytometry analysis of Histone H3K79me2 was performed on Jurkat labeled with Anti-Histone H3K79me2 Rabbit Polyclonal Antibody (Product# 720079, 2-4 ug/ 1M cells) or with rabbit isotype control and detected with Goat anti-Rabbit IgG (H+L) Superclonal™ Secondary Antibody, (Alexa Fluor® 488 conjugate, Product # A27034, 0.4 ug/ml, 1:2500) as represented by the red and pink histograms respectively. The purple histogram represents unstained control cells and the green histogram represents no-primary-antibody control. A representative of 10,000 cells were acquired and analyzed for each sample using an Attune® Acoustic Focusing Cytometer (4468770).
Flow Cytometry analysis of Histone H3K79me2 was performed on Jurkat labeled with Anti-Histone H3K79me2 Rabbit Polyclonal Antibody (Product# 720079, 2-4 ug/ 1M cells) or with rabbit isotype control and detected with Goat anti-Rabbit IgG (H+L) Superclonal™ Secondary Antibody, (Alexa Fluor® 488 conjugate, Product # A27034, 0.4 ug/ml, 1:2500) as represented by the red and pink histograms respectively. The purple histogram represents unstained control cells and the green histogram represents no-primary-antibody control. A representative of 10,000 cells were acquired and analyzed for each sample using an Attune® Acoustic Focusing Cytometer (4468770).
Related Products
Related Images