Meprin beta Subunit / MEP1B Antibody (PA5-47474) in IHC (F)
Immunohistochemical analysis of Meprin β Subunit/MEP1B was detected in perfusion fixed frozen sections of mouse intestine using 1 µg/mL Goat Anti-mouse Meprin β Subunit/MEP1B Antigen Affinity-purified Polyclonal Antibody (Product # PA5-47474) overnight at 4 °C. Tissue was stained with the Anti-Goat HRP-DAB (brown and counterstained with hematoxylin (blue). Specific labeling was localized to the brush border of intestinal villi.
Immunohistochemical analysis of Meprin β Subunit/MEP1B was detected in perfusion fixed frozen sections of mouse intestine using 1 µg/mL Goat Anti-mouse Meprin β Subunit/MEP1B Antigen Affinity-purified Polyclonal Antibody (Product # PA5-47474) overnight at 4 °C. Tissue was stained with the Anti-Goat HRP-DAB (brown and counterstained with hematoxylin (blue). Specific labeling was localized to the brush border of intestinal villi.
Related Products
Related Images