Search
Immunohistochemistry was performed on mouse lymph node tissues. Samples were probed with a Monoclonal Mannose Receptor / CD206 anditbody (Product #MA5-16869) and stained with Immunoperoxidase, followed by horseradish peroxidase conjugated Goat anti Rat IgG antibody.