Search
CHO cells expressing hERG potassium channels were preloaded with the indicated reagent using the supplier protocol provided and stimulated with a solution containing 2mM thallium and 10 mM potassium to activate voltage gated potassium channels. The cells were exposed to different concentrations of the potassium channel blocker E-4031 and then detected using a Hamamatsu FDSS 6000 instrument. The FluxOR II Green Potassium Ion Channel Assay displayed a larger dynamic range of detection compared to the first-generation FluxOR assay or the Molecular Devices FLIPR™ assay, facilitating the analysis of a wider range of potassium channels, including low-expressed or weakly conducting channels.