p53 Antibody (AHO0152) in IP-MS
IP-MS enrichment of TP53 (LFQ intensity): TP53 was enriched 478-fold from BT549 lysate compared to background proteins, using the optimized IP-MS workflow with Pierce MS-Compatible Magnetic IP Kit protein A/G (Product # 90409) and TP53 antibody (Product # AHO0152). STRING database was used to identify the protein interactor list. See more information on IP-MS verification of antibody selectivity.
IP-MS enrichment of TP53 (LFQ intensity): TP53 was enriched 478-fold from BT549 lysate compared to background proteins, using the optimized IP-MS workflow with Pierce MS-Compatible Magnetic IP Kit protein A/G (Product # 90409) and TP53 antibody (Product # AHO0152). STRING database was used to identify the protein interactor list. See more information on IP-MS verification of antibody selectivity.
Related Products
Related Images