Search
IP-MS enrichment of HRAS (LFQ intensity): HRAS was enriched 270-fold from HCT116 lysate compared to background proteins, using the optimized IP-MS workflow with Pierce MS-Compatible Magnetic IP Kit protein A/G (Part No. 90409) and HRAS antibody (Part No. 33-7200). The STRING database (www.string-db.org) was used to identify the protein interactor list. See more information on IP-MS verification of antibody selectivity.