Search
Western blot analysis was performed on whole cell extracts (30 µg lysate) of A549 (Lane 1) and Hep G2 (Lane 2). The blots were probed with Anti-Calyculin Polyclonal Antibody (Product ## 720326, 1 µg/ml) and detected by chemiluminescence using Goat anti-Rabbit IgG (H+L) Superclonal™ Secondary Antibody, HRP (Product ## A27036) at dilutions 1 µg/ml (Fig. 1), 0.5 ug/ml (Fig. 2) and 0.05 µg/ml (Fig. 3). A 30 kDa band corresponding to Calcyclin was observed. Known quantity of protein samples were electrophoresed using Novex® NuPAGE®12 % Bis-Tris gel (Product ## NP0342BOX), XCell SureLock™ Electrophoresis System (Product ## EI0002) and Novex® Sharp Pre-Stained Protein Standard (Product ## LC5800). Resolved proteins were then transferred onto a nitrocellulose membrane with iBlot® 2 Dry Blotting System (Product ## IB21001). The membrane was probed with the relevant primary and secondary antibody after blocking with 5 % skimmed milk. Chemiluminescent detection was performed using Novex® ECL Chemiluminescent Substrate Reagent Kit (Product ## WP20005).