Search
The biological activity of EGF was tested in culture by measuring its ability to stimulate proliferation of TF-1 human erythroleukemic cells. After a 72-hour treatment with EGF, cells were incubated for 2 hours with WST8 and read at 570/630 nm.