Kit specificity.
The IL-12 Human ELISA kit quantifies natural and recombinant human IL-12 p35/p40 heterodimer, also known as IL-12p70, and also detects the free p40 subunit.
The IL-12 Human ELISA kit quantifies natural and recombinant human IL-12 p35/p40 heterodimer, also known as IL-12p70, and also detects the free p40 subunit.
Related Products
Related Images