EfficientFeed™ C+ supplement super-concentration
The same package of EfficientFeed™ C+ supplement can be reconstituted at up to 2X the original concentration by simply reducing the total amount of water used (see user manual for more information on reconstitution).
The same package of EfficientFeed™ C+ supplement can be reconstituted at up to 2X the original concentration by simply reducing the total amount of water used (see user manual for more information on reconstitution).
Related Products
Related Images