Specifications of Pierce Centrifuge Columns
All columns are made from low protein binding polypropylene for compatibility with protein purification, and fit into standard centrifuge tubes for use in any centrifuge.
All columns are made from low protein binding polypropylene for compatibility with protein purification, and fit into standard centrifuge tubes for use in any centrifuge.
Related Products
Related Images