Specifications of Pierce Spin Cups and Columns
All columns are made from low protein binding polypropylene for compatibility with protein purification, and fit into standard microcentrifuge tubes for use in most microcentrifuges.
All columns are made from low protein binding polypropylene for compatibility with protein purification, and fit into standard microcentrifuge tubes for use in most microcentrifuges.
Related Products
Related Images