Microtubules of a fixed bovine pulmonary artery endothelial cell. Anti–bovine a-tubulin, Rhodamine Red™-X goat anti–mouse IgG (H+L) conjugate and DAPI.
Microtubules of a fixed bovine pulmonary artery endothelial cell labeled with mouse monoclonal anti–a-tubulin antibody (Cat. no. A11126), and detected with Rhodamine Red™-X goat anti–mouse IgG (Cat. no. R6393). The nucleus was counterstained with DAPI (Cat. no. D1306, D3571, D21490). The multiple-exposure image was acquired using bandpass filters appropriate for Texas Red® dye and DAPI.
Microtubules of a fixed bovine pulmonary artery endothelial cell labeled with mouse monoclonal anti–a-tubulin antibody (Cat. no. A11126), and detected with Rhodamine Red™-X goat anti–mouse IgG (Cat. no. R6393). The nucleus was counterstained with DAPI (Cat. no. D1306, D3571, D21490). The multiple-exposure image was acquired using bandpass filters appropriate for Texas Red® dye and DAPI.
Related Products
Related Images