Search
A mixed population of Bacillus cereus and Pseudomonas aeruginosa stained with the dye mixture provided in our LIVE BacLight™ Gram Stain Kit (Cat. No. L7005). When live bacteria are incubated with this kit's cell-permeant nucleic acid stains, gram-positive organisms fluoresce orange and gram-negative organisms fluoresce green. The bacteria were photographed through an Omega Optical triple bandpass filter set.
Cyanobacteria. BODIPY® FL, DAPI and CellTracker™ Green BODIPY®. Go ›
Bifidobacterium sp. bacteria stained with the ViaGram™ Red+ Bacterial Gram Stain and Viability Kit. Go ›
BPAE cells fixed and permeabilized using the Image-iT® Fixation/Permeabilization Kit. Go ›
BPAE cells fixed and permeabilized using the Image-iT® Fixation/Permeabilization Kit. Go ›
Live cell imaging with CellLight™ reagents. Go ›
Live cells transduced with Organelle Lights™ or Cellular Lights™ reagents. Go ›
CD335 (NKp46) Antibody (63335182) in RE Go ›
CD223 (LAG-3) Antibody (56223942) in TM Go ›