Bifidobacterium sp.
Bifidobacterium sp. stained with the reagents in the LIVE/DEAD® BacLight™ Bacterial Viability Kit (Cat. No. L7007, L7012). When incubated with the SYTO® 9 stain and propidium iodide nucleic acid stain provided in this kit, live bacteria with intact cell membranes fluoresce green and dead bacteria with compromised membranes fluoresce red.
<i>Bifidobacterium sp.</i> stained with the reagents in the LIVE/DEAD® <i>Bac</i>Light™ Bacterial Viability Kit (Cat. No. L7007, L7012). When incubated with the SYTO® 9 stain and propidium iodide nucleic acid stain provided in this kit, live bacteria with intact cell membranes fluoresce green and dead bacteria with compromised membranes fluoresce red.
Related Products
Related Images