Search
1 kb, 4 kb, or 9 kb templates were used in the MEGAscript™ Kit or in another manufacturer's in-vitro transcription kit according to protocol. A portion of each reaction (5 µL) was loaded onto a 1% formaldehyde gel and stained with ethidium bromide.