Search
Immunofluorescence analysis of neurons using anti-MAP2 antibody (MA5-12823). GABAergic precursor cells derived from H9 embryonic stem cells were differentiated for 7 days and stained with antibodies against MAP2 (Cat. No. MA5-12823 at 1:200) followed by Alexa Fluor 488 goat anti-rabbit (Cat. No. A21206, green). Nuclear DNA was stained with DAPI (blue) in the merged image (right panel).