Search
Pseudocolored micrograph of a single living smooth muscle cell labeled with a reduced form of rhod-2 AM (Cat. no. R1244, R1245MP), dihydrorhod-2. Images were acquired at focal planes spaced at 0.25 µm intervals and then processed using a constrained iterative deconvolution algorithm. This image shows that the rhod-2 fluorescence primarily arises from the mitochondria. The image was contributed by Fredric S. Fay, Program in Molecular Medicine, University of Massachusetts Medical Center.
CD335 (NKp46) Antibody (63335182) in RE Go ›
CD223 (LAG-3) Antibody (56223942) in TM Go ›
REF-52 fibroblasts. Cyclic AMP Fluorosensor (FlCRhR) and fura-2 AM Go ›
Pseudocolored images of changes in intracellular free Ca2+ in AtT-20/D16v-F2 cells, monitored at 9 sec intervals with fluo-4, AM (F14201, F14202, F14217, F23917). Go ›
Live cell imaging with CellLight™ reagents. Go ›
Live cells transduced with Organelle Lights™ or Cellular Lights™ reagents. Go ›