Search
Live CRE BAG 2 cells loaded with the beta-galactosidase substrate from the DetectaGene Green CMFDG lacZ Gene Expression Kit (Cat. No. D2920). The substrate was loaded into cells with the aid of the Influx™ pinocytic cell-loading reagent (Cat. No. I14402), a component of the DetectaGene™ Green Kit. The cells were then stained with LysoTracker® Red DND-99 (Cat. No. L7528) to differentiate lysosomal from cytoplasmic localization of the green-fluorescent CMFDG product, as well as to illustrate the selective rupture of only the Influx™ reagent–induced pinocytic vesicles. The double-exposure image was acquired using bandpass filters appropriate for fluorescein and Texas Red® dye.
Zymographic detection of ß-D-glucuronidase activity. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). Go ›
CD335 (NKp46) Antibody (63335182) in RE Go ›
CD223 (LAG-3) Antibody (56223942) in TM Go ›
Live cell imaging with CellLight™ reagents. Go ›
Bacteriophage T4 replication proteins stained with SYPRO® Orange Protein Gel Stain. Go ›
lacZ-negative (NIH 3T3) and lacZ-positive (CRE BAG 2) cells. Go ›
Adherent CRE BAG 2 cells. Alexa Fluor 594 hydrazide and Influx pinocytic cell-loading reagent. Go ›
Live cells transduced with Organelle Lights™ or Cellular Lights™ reagents. Go ›
Live cells transduced with Organelle Lights™ or Cellular Lights™ reagents. Go ›