Search
Horseradish peroxidase (HRP)–filled reticulospinal neurons of the hindbrain of a whole-mount zebrafish larva were detected using green-fluorescent Alexa Fluor® 488 tyramide (TSA Kit #2, Cat. no. T20912). The spinal cord was labeled by transection in the presence of HRP (1 mg/mL). After a two-hour incubation, the specimen was fixed and the HRP was visualized using Alexa Fluor® 488 tyramide. This figure represents a projection (performed with AutoQuant Imaging, Inc., software) of a stack of 30 images.
Simultaneous detection of expression of five genes in a whole mount Drosophila embryo by fluorescence in situ hybridization (FISH) with five RNA probes Go ›
Immunocytochemistry using Alpha-Tubulin Monoclonal Antibody, Mouse Go ›
Developing Drosophila embryo Go ›
Cytoskeleton of a mixed population of granule neurons and glial cells Go ›
Zymographic detection of ß-D-glucuronidase activity. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). Go ›
CD335 (NKp46) Antibody (63335182) in RE Go ›
CD223 (LAG-3) Antibody (56223942) in TM Go ›