Neuronal cells in a 22-hour zebrafish embryo.
Neuronal cells in a 22-hour zebrafish embryo were identified with anti–HuC/HuD mouse monoclonal antibody (Cat. No. A21271) and visualized with red-orange–fluorescent Alexa Fluor® 568 goat anti–mouse IgG antibody (Cat. No. A11004). The nuclei were stained with blue-fluorescent DAPI (Cat. No. D1306, D3571, D21490).
Related Products
Related Images
A prometaphase muntjac skin fibroblast stained with Alexa Fluor® 350 phalloidin, an anti–a-tubulin antibody and an anti–cdc6 peptide antibody. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
NF2 Antibody (711496) in KD Go ›
Phospho-VEGF Receptor 2 (Tyr1214) Antibody (44-1052) in ARRAY Go ›
Increase in accuracy and precision using the Qubit® Go ›
Performance of the Qubit® dsDNA HS Assay Go ›
Developing Drosophila embryo Go ›
Cytoskeleton of a mixed population of granule neurons and glial cells Go ›