Search
The QuantStudio 3 and 5 real-time PCR system workflow is supported by the following software applications.
This desktop instrument operating software enables users to operate QuantStudio 3 and 5 instruments. Customers who need to operate the QuantStudio 3/5 system within the desktop software or to access the Securities, Audits, and e-Signature module for the QuantStudio 5 system must use the Design and Analysis v1.x series.
Design & Analysis 2 software enables users to set up run templates, open completed run files to analyze data, and review results in an integrated and streamlined fashion. The software provides a suite of secondary analysis modules for specific qPCR applications.
While DA2 software can be run with Security, Audit, and e-Signature (SAE) enabled, this feature does not apply to the QuantStudio 3/5 systems.
The latest Design and Analysis software allows users to enjoy all the same features as Design & Analysis 2 (DA2) software while offering additional features and enhancements. Users can now access the software in any one of three deployment options: classic desktop experience (available in Windows or MAC), cloud flexibility, and on-premises server-client configuration for multi-user access via the security, audit, and e-signature (SAE) enabled environment*. Utilize multiplate analysis capabilities with both genotyping and copy number variation analysis modules. Diomni Design and Analysis (RUO) Software v3.0 is the feature-rich solution that connects all the steps of your qPCR workflows into a single software experience.
*SAE is not available with the desktop version of software. SAE is only available with the on-premises version of software.
If you have purchased a QuantStudio 3 or 5 real-time PCR system, you may download the system software above.
The software is available in a Thermo Fisher Connect folder. To download the software:
The change of firmware and software version may impact validated workflows. You must continue to follow current Instructions for Use (IFU) for validated workflows and/or perform all validations as required by your organization’s standard operating procedures before performing a software upgrade.
For Research Use Only. Not for use in diagnostic procedures.