Search
Optimize your experiments to get the best results. We’ve compiled a detailed knowledge base of the top tips and tricks to meet your research needs.
View the relevant questions below:
Having problems with your experiment? Visit our
Our Original Bolt™ Bis-Tris Plus Mini gels (Cat. No. BGxxxxxBOX, discontinued as of December 31, 2014) can only be run in the Bolt™ Mini Gel Tank (discontinued as of December 31, 2014, and will be offered until inventory is depleted).
Our New Bolt™ Bis-Tris Plus Mini gels (Cat. No. NWxxxxxBOX), as well as our Novex™ Mini gels and NuPAGE™ Mini gels can be run using the Mini Gel Tank, or XCell SureLock™ Mini-Cell. To run these gels using the Bolt™ Mini Gel Tank (discontinued as of December 31, 2014), upgrading of the tank is necessary by replacing the black 10.5 cm cassette clamp cam handles with gray 10 cm cassette clamp cam handles (Cat. No. A26732, Cassette Clamp Cam Handle Set). Instructions for replacement of the cam handles can be found here or in this video.
Our Midi gels can be run using the XCell4 SureLock™ Midi-Cell.
Yes, the 10 cm x 10 cm x 7 mm cassette Precise Tris-Glycine gels are compatible with the XCell SureLock™ Mini Cell and the Mini Gel Tank.
The Precise™ Tris-HEPES gels are compatible with the XCell SureLock™ Mini Cell or Mini Gel Tank when used with adaptor plates.
Note: Two adaptor plates are required when running just one gel and one adaptor plate is required when running two gels using the XCell SureLock™ Mini Cell or Mini Gel Tank.
We used to offer the XCell6™ MultiGel unit for simultaneous running of up to 6 mini gels, but it has been discontinued.
The XCell SureLock™ Mini-Cell includes the lid, gel tension wedge, lower buffer chamber, buffer core, buffer dam, and gel knife.
Our New Bolt™ Bis-Tris Plus Mini gels (Cat. No. NWxxxxxBOX), as well as our Novex™ Mini gels and NuPAGE™ Mini gels can be run using the XCell SureLock™ Mini-Cell.
The XCell II™ Mini-Cell has been discontinued and can be upgraded to an XCell SureLock™ Mini-Cell with the purchase of the SureLock™ Retrofit kit for XCell II™ Mini-Cell, Cat. No. EI0020 (consists of a SureLock™ mini-cell lid, gel tension wedge, and buffer dam).
We recommend washing the XCell SureLock™ Mini-Cell with a mild detergent and rinsing with deionized water after each use.
You can try the standard recommendation for cleaning which would be washing the unit with a mild detergent and rinsing with deionized water.
The XCell SureLock™ Mini-Cell is impervious to alcohol, but not compatible with chlorinated hydrocarbons (e.g., chloroform), aromatic hydrocarbons (e.g., toluene, benzene) or acetone.
Here are the replacement parts we offer for the XCell SureLock™ Mini-Cell:
Our Original Bolt™ Bis-Tris Plus gels (Cat. No. BGxxxxxBOX), discontinued as of December 31, 2014) do not fit in the XCell SureLock™ Mini-Cell. They need to be run in the Bolt™ Mini Gel Tank (discontinued as of December 31, 2014). Our New Bolt™ Bis-Tris Plus Mini gels (Cat. No. NWxxxxxBOX) can be run using the XCell SureLock™ Mini-Cell (or Mini Gel Tank, Cat. No. A25977).
The Mini Gel Tank includes the Mini Gel Electrophoresis Tank, Mini Gel Tank Lid, Cassette Clamp (Right), 10 cm, Cassette Clamp (Left), 10 cm, Mini Gel Tank Base, Novex™ Power Supply Adapters, and Gel Knife.
Our New Bolt™ Bis-Tris Plus Mini gels (Cat. No. NWxxxxxBOX), as well as our Novex™ Mini gels and NuPAGE™ Mini gels can be run using the Mini Gel Tank. Please note that our original Bolt™ Bis-Tris Plus Mini gels (Cat. No. BGxxxxxBOX, discontinued as of December 31, 2014) can only be run in the Bolt™ Mini Gel Tank (discontinued as of December 31, 2014, and will be offered until inventory is depleted).
Please note that the Bolt™ Mini Gel Tank has been discontinued as of December 31, 2014 and will be available for purchase until inventory is depleted. The Mini Gel Tank is an improved version of the Bolt™ Mini Gel Tank with enhancements in design allowing for:
Please check out this video for operating the Mini Gel Tank.
The Mini Gel Tank is not compatible with chlorinated hydrocarbons (e.g., chloroform), aromatic hydrocarbons (e.g., toluene, benzene) or acetone.
Here are our recommendations:
| Replacement part | Cat. No. |
| Gel Runner Tank | B4478641 |
| Mini Gel Tank Lid | A25944 |
| Mini Gel Tank Base | A25950 |
| Cassette Clamp, left | A25946 |
| Cassette Clamp, right | A25945 |
| Cassette Clamp Cam Handle Set | A26732 |
Our Original Bolt™ Bis-Tris Plus Mini gels (Cat. No. BGxxxxxBOX, discontinued as of December 31, 2014) can only be run in the Bolt™ Mini Gel Tank (discontinued as of December 31, 2014, and will be offered until inventory is depleted).
To run our new Bolt™ Bis-Tris Plus Mini gels (Cat. No. NWxxxxxBOX), as well as our Novex™ Mini gels and NuPAGE™ Mini gels using the Bolt™ Mini Gel Tank, upgrading of the tank is necessary by replacing the black 10.5 cm cassette clamp cam handles with gray 10 cm cassette clamp cam handles (Cat. No. A26732, Cassette Clamp Cam Handle Set). Instructions for replacement of the cam handles can be found in this video. Additional resources can be found here.
To run Mini gels with 10 cm gel cassettes using the Bolt™ Mini Gel Tank (without replacement of 10.5 cm cassette clamp cam handles with 10 cm cassette clamp cam handles), please use the instructions provided on Page 22 of the manual.
The Bolt™ Mini Gel Tank has been discontinued as of December 31, 2014 and will be available for purchase until inventory is depleted. The Gel Runner Tank (Cat. No. B4478641) is still available. The Cassette Clamp, left (Cat. No. B4478593), Cassette Clamp, right (Cat. No. B4478592), Bolt™ Tank Base (Cat. No. B4478640), and Bolt™ Tank Lid (Cat. No. B4478591) have been discontinued as of Dec 31, 2014 and will be available for purchase until inventory is depleted. The alternative SKUs are Cassette Clamp, left (Cat. No. A25946), Cassette Clamp, right (Cat. No. A25945), Bolt™ Tank Base (Cat. No. A25950), and Bolt™ Tank Lid (Cat. No. A25944)
To run our new Bolt™ Bis-Tris Plus Mini gels (Cat. No. NWxxxxxBOX), as well as our Novex™ Mini gels and NuPAGE™ Mini gels using the Bolt™ Mini Gel Tank, upgrading of the tank is necessary by replacing the black 10.5 cm cassette clamp cam handles with gray 10 cm cassette clamp cam handles (Cat. No. A26732, Cassette Clamp Cam Handle Set). Instructions for replacement of the cam handles can be found in this video. Additional resources can be found here.
The Bolt™ Mini Gel Tank is impervious to alcohol, but not compatible with chlorinated hydrocarbons (e.g., chloroform), aromatic hydrocarbons (e.g., toluene, benzene) or acetone.
Note: The Bolt™ Mini Gel Tank has been discontinued as of December 31, 2014, and will be offered until inventory is depleted.
We recommend the following:
Note: The Bolt™ Mini Gel Tank has been discontinued as of December 31, 2014, and will be offered until inventory is depleted.
We offer the PowerEase™ 90W Power supply that is compatible with the Bolt™ Mini Gel Tank. The PowerEase™ 500 Power Supply (discontinued as of December 31, 2013) is also compatible with the Bolt™ Mini Gel Tank). Novex™ Power Supply Adapters are supplied with the tank for power supplies that are not designed for use with covered or non-retractable power leads.
Note: The Bolt™ Mini Gel Tank has been discontinued as of December 31, 2014, and will be offered until inventory is depleted.
To run Mini gels with 10 cm gel cassettes using a Bolt™ Mini Gel Tank (without replacement of 10.5 cm cassette clamp cam handles with 10 cm cassette clamp cam handles), please use the instructions provided on Page 22 of the manual.
For optimal results, to run 10 cm cassette Mini gels with a Bolt™ Mini Gel Tank, you should replace the black 10.5 cm cassette clamp cam handles on the Bolt™ Mini Gel Tank with gray 10 cm cassette clamp cam handles (Cat. No. A26732). Instructions for replacement of the cam handles can be found on Page 20 of the manual or in this video. Additional resources can be found here.
Note: The Bolt™ Mini Gel Tank has been discontinued as of December 31, 2014, and will be offered until inventory is depleted.
The Bolt™ Mini Gel Tank has been discontinued as of December 31, 2014, and will be offered until inventory is depleted. To upgrade the Bolt™ Mini Gel Tank to run the new Bolt™ Bis-Tris Plus gels as well as other Novex™ Mini gels, you have to replace the black 10.5 cm cassette clamp cam handles with gray 10 cm cassette clamp cam handles (Cat. No. A26732). Instructions for replacement of the cam handles can be found in this video. Additional resources can be found here.
Please note that the Bolt™ Mini Gel Tank has been discontinued as of December 31, 2014 and will be available for purchase until inventory is depleted. The Mini Gel Tank is an improved version of the Bolt™ Mini Gel Tank with enhancements in design allowing for:
Please check out this video for operating the Mini Gel Tank.
The XCell4 SureLock™ Midi-Cell includes the lower buffer chamber, lid with cables, two buffer cores, buffer dam, displacement dam, SureLock™ assembly, and gel knife.
Our Midi gels can be run using the XCell4 SureLock™ Midi-Cell.
Yes, the XCell4 SureLock™ Midi-Cell is designed to perform simultaneous vertical electrophoresis of 1–4 midi-gels. Use the XCell4 Displacement Dam when you wish to run less than 3 gels in the XCell4 SureLock™ Midi-Cell. The Displacement Dam is placed in the Lower Buffer Chamber instead of a second Buffer Core. Do not add any buffer or water to the Displacement Dam. However, if water or buffer is accidentally added to the Displacement Dam, it does not affect the electrophoresis run.
No. While the NuPAGE™ Novex™ Midi Gels are compatible with the Bio-Rad™ Criterion system (when the specific adapter is used), the Bio-Rad™ Criterion gels will not work with the XCell4 SureLock™ Midi-Cell.
Note: Our Mini gels are not compatible with the Bio-Rad™ Mini-PROTEAN™ apparatus.
Here are the replacement parts we offer for the XCell4 SureLock™ Midi-Cell. They can only be ordered by phone:
| Replacement Part | Cat. No. |
| XCell4 SureLock™ Midi-Cell Lid | WR0101 |
| XCell4 SureLock™ Midi-Cell Lower Buffer Chamber | WR0102 |
| XCell4 SureLock™ Midi-Cell Displacement Dam | WR0106 |
| XCell SureLock™ Upgrade kit (contains 1 cam, 1 buffer dam and 2 cores) | WR0112 |
The XCell4 SureLock™ Midi-Cell is impervious to alcohol, but not compatible with chlorinated hydrocarbons (e.g., chloroform), aromatic hydrocarbons (e.g., toluene, benzene) or acetone.
Use the XCell4 Displacement Dam when you wish to run less than 3 gels in the XCell4 SureLock™ Midi-Cell. The Displacement Dam is placed in the Lower Buffer Chamber instead of a second Buffer Core. Do not add any buffer or water to the Displacement Dam. However, if water or buffer is accidentally added to the Displacement Dam, it does not affect the electrophoresis run.
The XCell4 SureLock™ Assembly or CAM is similar to a gel tension wedge. The positive locking action of the XCell SureLock™ Assembly ensures trouble-free, leak-free electrophoresis of protein gels. Move the lever on the XCell SureLock™ Assembly to a locked position to generate an even horizontal force which seals the gel cassette/Buffer Core assembly firmly into position in the Lower Buffer Chamber. The locked and unlocked positions are marked on the XCell SureLock™ Assembly for easy, intuitive assembly.
We do not recommend using the gel tension wedge that is supplied with the XCell SureLock™ Mini-Cell for the XCell4 SureLock™ Midi-Cell as it is specifically designed for the Mini-Cell and will not fit correctly in the Midi-Cell.
The XCell4 SureLock™ Midi-Cell should be used with an external DC power supply designed for electrophoresis. This power supply must:
If you would like to simultaneously run 4 gels using the XCell4 SureLock™ Midi-Cell, we recommend using the ZOOM™ Dual Power Supply. The Low Voltage/High Current outlets can handle the voltage and current of 4 NuPAGE™ Bis-Tris gels (Voltage 200 V, Current ~800 mA).
The PowerEase™ 90W Power Supply and PowerEase™ 500 Power Supply (discontinued as of December 31, 2013) can be used with the XCell4 SureLock™ Midi-Cell but the current limit of 500 mA will be reached with 2 gels (2 x ~200 mA max current at 200V). If 4 gels are run in the XCell4 SureLock™ Midi-Cell with these power supplies, the current will be maxed out and the run will slow down. It will take approximately twice as long to run the gels.
Note: To allow connection of the covered power leads of the XCell4 SureLock™ Midi-Cell with certain power supplies that are not designed for use with covered or non-retractable power leads, use Novex™ Power Supply Adapters available separately (Cat. No. ZA10001).
We recommend washing the unit with a mild detergent and rinsing with deionized water after each use.
The PowerEase™ 300W Power Supply is a fully programmable, microprocessor-controlled power supply designed for high-throughput gel electrophoresis. It has a straight-forward, intuitive interphase and includes the following features:
The PowerEase™ 300W Power Supply accommodates the running and transferring of 8 Mini gels, or 16 Midi gels.
The PowerEase™ 300W Power Supply includes the following components:
Note: Please use the power cord approved for your country/region. North America customers should use the Type B power cord; Japanese customers should use the JIS 8303 power cord; Chinese customers should use the GB 15934-2008 power cord; Australia/New Zealand customers should use the Type I power cord.
The product specifications for the PowerEase™ 300W Power Supply are listed on Page 18 of the manual.
For best results, we do not recommend using the PowerEase™ 300W Power Supply at its maximum electrical load limits. Variations in buffer conditions can result in exceeding the power supply’s maximum voltage, current, or power output capacity and produce undesirable variations in performance.
When using the PowerEase™ 300W Power Supply with 1–2 proteins gels or blots, we recommend using the Constant Voltage mode for optimal performance.
The PowerEase™ 300W Power Supply is capable of operating at limiting voltage, limiting current, or limiting power. However, we recommend operating it at limiting voltage for most applications as it provides the following advantages:
Note:
With the current limiting setting, the voltage will increase as resistance increases to satisfy Ohm’s law (V=IR). If no voltage limit is set and a local fault condition occurs, such as a poor connection, very high local resistance may cause the voltage to increase to the maximum capacity of the power supply. This may lead to local overheating and damage to the electrophoresis cell or create unsafe conditions. When operating under constant current conditions, we recommend setting a voltage limit on the power supply at or slightly above the maximum expected voltage.
Power is a function of voltage and current P=IV. For a typical gel run, the current will decrease causing the voltage to increase when run at constant power. The power limiting function may be used when running sequencing gels to remove the ammonium persulfate from the wells and to heat the gel to an optimal temperature for DNA separations.
The PowerEase™ 300W Power Supply requires no periodic maintenance with the exception of an occasional dry wipe-down of the instrument.
For detailed instructions on replacing the fuse on the PowerEase™ 300W Power Supply, please refer to Page 19 of the manual.
During NativePAGE™ electrophoresis, it is common for the current to drop below 1 mA. The PowerEase™ 300W Power Supply will register this as a “No Load” error and automatically shut off, resulting in the stopping of the gel run. Since there is no way to disable/turn off the “Load Check” feature in the PowerEase™ 300W Power Supply, we do not recommend using it for Native-PAGE electrophoresis.
The PowerEase™ 90W Power Supply is a microprocessor-controlled power supply designed specifically for mini-gel electrophoresis. The straight-forward, intuitive interface makes the powering of DNA and protein gel runs a simple and easy process. In addition, the PowerEase™ 90W Power Supply features:
The PowerEase™ 90W Power Supply accommodates the running of 2 Mini gels, or 2 Midi gels.
The PowerEase™ 90W Power Supply includes the following components:
Note: Please use the power cord approved for your country/region. North America customers should use the Type B power cord; Japanese customers should use the JIS 8303 power cord; Chinese customers should use the GB 15934-2008 power cord; Australia/New Zealand customers should use the Type I power cord.
The product specifications for the PowerEase™ 90W Power Supply are listed on Page 16 of the manual.
For best results, we do not recommend using the PowerEase™ 90W Power Supply at its maximum electrical load limits. Variations in buffer conditions can result in exceeding the power supply’s maximum voltage, current, or power output capacity and produce undesirable variations in performance.
The PowerEase™ 90W Power Supply is capable of operating at limiting voltage, or limiting current. However, we recommend operating it at limiting voltage for most applications as it provides the following advantages:
Note: With the current limiting setting, the voltage will increase as resistance increases to satisfy Ohm’s law (V=IR). If no voltage limit is set and a local fault condition occurs, such as a poor connection, very high local resistance may cause the voltage to increase to the maximum capacity of the power supply. This may lead to local overheating and damage to the electrophoresis cell or create unsafe conditions. When operating under constant current conditions, we recommend setting a voltage limit on the power supply at or slightly above the maximum expected voltage.
The PowerEase™ 90W Power Supply requires no periodic maintenance with the exception of an occasional dry wipe-down of the instrument.
For detailed instructions on replacing the fuse on the PowerEase™ 90W Power Supply, please refer to Page 17 of the manual.
The PowerEase™ 500 Power Supply has been discontinued. Our newer alternatives are the PowerEase™ 300W Power Supply and PowerEase™ 90W Power Supply.
Yes, however, keep in mind that the LCD screen will fade away in the cold room. The PowerEase™ would still be working and this will be indicated by the green and red light. The screen should come back on when returned to room temp.
Note: The PowerEase™ 500 Power Supply has been discontinued as of December 31, 2013 and our newer alternatives are the PowerEase™ 300W Power Supply and PowerEase™ 90W Power Supply.
The PowerEase™ 500 Power Supply can be operated under a variety of conditions including constant voltage, constant current and constant power. The default setting is for constant voltage. We recommend running our precast gels at constant voltage for the following reasons:
If you would like to use the PowerEase™ 500 Power Supply to run a gel at constant current, this can be accomplished by setting a voltage limit (at or slightly above the maximum voltage expected to be reached during the run) and then setting the current to the desired level. In this way, the device will try to reach the set voltage. However, when it reaches the threshold current, the current will plateau and the run will switch to constant current (the limiting factor). If no voltage limit is set and a local fault condition occurs, such as a poor connection, very high local resistance may cause the voltage to increase to the maximum of the power supply. This can lead to local overheating and damage to the electrophoresis unit and or gel(s) or create unsafe conditions.
Note: The PowerEase™ 500 Power Supply has been discontinued as of December 31, 2013 and our newer alternatives are the PowerEase™ 300W Power Supply and PowerEase™ 90W Power Supply.
The PowerEase™ 500 Power Supply can be run under a variety of conditions including constant voltage, constant current, and constant power. The default setting is for constant voltage. We recommend running our precast gels at constant voltage for the following reasons:
If you want to use the PowerEase™ 500 Power Supply to run a gel at constant power, this can be accomplished by setting a voltage limit (at or slightly above the maximum voltage expected to be reached during the run) and then setting the power to the desired level. If no voltage limit is set and a local fault condition occurs, such as a poor connection, very high local resistance may cause a high proportion of the total heat to be generated over a small distance, damaging the electrophoresis unit and/or gel(s).
Note: The PowerEase™ 500 Power Supply has been discontinued as of December 31, 2013 and our newer alternatives are the PowerEase™ 300W Power Supply and PowerEase™ 90W Power Supply.
Other than an occasional wipe-down of the instrument to remove salt deposits, which may have built up over time, the PowerEase™ 500 Power Supply does not require a periodic maintenance program.
Note: The PowerEase™ 500 Power Supply has been discontinued as of December 31, 2013 and our newer alternatives are the PowerEase™ 300W Power Supply and PowerEase™ 90W Power Supply.
Using the PowerEase™ 500 Power Supply, the number of Tris-Glycine or NuPAGE™ gels that can be run simultaneously is shown below:
Tris-Glycine gels: 10 gels i.e., 5 XCell SureLock™ Mini-Cells
NuPAGE™ Bis-Tris gels: 2 gels i.e., 1 XCell SureLock™ Mini-Cell
NuPAGE™ Tris-Acetate gels: 6 gels i.e., 3 XCell SureLock™ Mini-Cells
Note: The PowerEase™ 500 Power Supply has been discontinued as of December 31, 2013 and our newer alternatives are the PowerEase™ 300W Power Supply and PowerEase™ 90W Power Supply.
Linear mode is a feature of the PowerEase™ 500 Power Supply that has not been described in the manual. The linear mode can be activated from the 'Edit' screen (where you would change settings for time, voltage, power, current). While on the “Edit” screen, press the left and right arrows simultaneously. This will activate another screen with a three choice menu where the third choice is linear mode. Move the cursor down to 'Linear mode' and turn on Linear mode by using the up/down arrows.
Note: The PowerEase™ 500 Power Supply has been discontinued as of December 31, 2013 and our newer alternatives are the PowerEase™ 300W Power Supply and PowerEase™ 90W Power Supply.
| Gel type | Time (hours:mins) | Voltage | Current | Wattage |
| Tris Glycine | 1:30 | 125 | 35 | 5 |
| NuPAGE™ | 0:35 | 200 | 120 | 25 |
| IEF | 1:00 | 100 | 18 | 2 |
| 1:00 | 200 | 18 | 3.5 | |
| 0:30 | 500 | 18 | 9 | |
| Western blot | 1:30 | 25 | 125 | 17 |
| NuPAGE™ western | 1:00 | 25 | 160 | 17 |
| Tricine | 1:15 | 15 | 90 | 12.5 |
Note: The PowerEase™ 500 Power Supply has been discontinued as of December 31, 2013 and our newer alternatives are the PowerEase™ 300W Power Supply and PowerEase™ 90W Power Supply.
The dimensions of the Gel Knife are: Handle 13 cm, Blade (9 cm (length) x 4.5 cm (width).
We do offer disposable, empty plastic cassettes and combs to provide the option of pouring your own Mini or Midi gels for use with the XCell SureLock™ Mini-Cell, Mini Gel Tank, or XCell4 SureLock™ Midi-Cell, respectively. All cassettes are sealed on three sides to prevent leaking. Since the cassettes are pre-sealed with the slot pre-taped, a bulky casting stand is not necessary. The empty cassettes and combs can be found here.
Yes, details below:
We offer Novex™ Gel Loading Tips for easy and accurate loading of samples. They are RNase/DNase-free and non-autoclavable. Three varieties are offered: standard round tips, flat gel tips, and Eppendorf™ round gel tips.
We offer the following systems for drying of protein gels:
The pore size of DryEase™ cellophane is 40 microns.
Yes, it is compatible. You need to put the gel in the enhancer solution, wash it and then proceed with the gel drying solution. For detection of signal using 32P, the cellophane is fine. For 35S, the upper layer of cellophane may need to be removed (which is possible) depending on the enhancer being used. For detection of 14C, there is no need to remove the cellophane as it will pass through.
The Gel-Dry™ Drying solution has a proprietary composition.
30% MeOH and 5% glycerol can be tried as a substitute for the Gel-Dry™ Drying solution.
We offer the following staining trays for staining protein gels:
StainEase™ Staining Trays (Cat. No. NI2400) for convenient and even staining of Mini and Midi gels. The staining tray assembly includes a perforated strainer insert that holds the gel and drains away the stain, a lid, and an outer container in which stains and solutions can be changed without having to handle the gel. Each tray easily fits up to 4 mini gels.
The ZOOM™ IEF Fractionator includes the following components:
The ZOOM™ IEF Fractionator should be used with an external DC power supply designed for electrophoresis and IEF applications. This power supply must:
Many power supplies automatically shut off when the current drops below 1 mA. You will need a power supply capable of overriding the low current shut-off feature. If the power supply has a current and power limiting capability, it is recommended to set a current limit at 2 mA and a power limit at 2 W.
The electrical leads of the ZOOM™ IEF Fractionator lid are recessed and may not fit into some power supply units. To allow connection of the ZOOM™ IEF Fractionator power leads with certain power supplies, use Novex™ Power Supply Adapters available separately (Cat. No. ZA10001).
Perform fractionation using the following conditions:
100 V for 20 minutes
200 V for 80 minutes
600 V for 80 minutes
The ZOOM™ IEF Fractionator is impervious to alcohol, but not compatible with chlorinated hydrocarbons (e.g., chloroform), aromatic hydrocarbons (e.g., toluene, benzene) or acetone.
We recommend washing the ZOOM™ IEF Fractionator with a mild detergent and rinsing with deionized water after each use. Do not wash lid with cables plugged into the power supply.
Here are the components of the ZOOM™ IPGRunner Mini-Cell:
We offer the ZOOM™ IPGRunner Retrofit kit for adapting the XCell SureLock™ Mini-Cell for two-dimensional (2D) electrophoresis.
The ZOOM™ IPGRunner Retrofit kit includes the ZOOM™ IPGRunner Buffer Core and Lid that are designed to be used with the lower buffer chamber of the XCell SureLock™ Mini-Cell for 2D electrophoresis.
We recommend washing the ZOOM™ IPGRunner™ Mini-Cell with a mild detergent and rinsing with deionized water after each use. Do not wash lid with cables plugged into the power supply.
The ZOOM™ IPGRunner Mini-Cell is impervious to alcohol, but not compatible with chlorinated hydrocarbons (e.g., chloroform), aromatic hydrocarbons (e.g., toluene, benzene) or acetone.
The ZOOM™ IPGRunner™ System should be used with an external DC power supply designed for electrophoresis applications. This power supply must:
Note: Many power supplies automatically shut off when the current drops below 1 mA. You will need a power supply capable of overriding the low current shut-off feature.
The electrical leads of the ZOOM™ IPGRunner™ Lid are recessed and may not fit into some power supply units. To allow connection of the ZOOM™ IPGRunner™ power leads with certain power supplies, use Novex™ Power Supply Adapters available separately (Cat. No. ZA10001).
The ZOOM™ Dual Power Supply includes the following components: ZOOM™ Dual Power Supply, extra fuse, and power cord.
We recommend running all applications that require 250 Volts or less on the low voltage/high current side of the ZOOM™ Dual Power Supply.
The ZOOM™ Dual Power supply requires no periodic maintenance program with the exception of an occasional dry wipe-down of the instrument.
For Research Use Only. Not for use in diagnostic procedures.