Search
Perform reliable, basic anion and cation analyses with the Thermo Scientific Dionex Aquion ion chromatography system. This simple, compact ion chromatography (IC) instrument, commonly used for water analysis, is designed for ease of use and straightforward operation for budget-limited labs. It supports electrolytic suppression and includes a vacuum degasser for inline degassing of eluents. It also offers fast start-up times and reliable, stable performance for demanding applications in environmental, food safety, and academic laboratories.
Conductivity detection
The inert, nonmetallic PEEK flow path of the Dionex Aquion IC system is designed and optimized for applications using conductivity detection. It ensures compatibility with corrosive eluents and IC analyses free from metal contamination. The high-performance conductivity cell is temperature controlled to protect against ambient temperature variations. Digital control of the cell offers a wide, dynamic range for detection.
Suppressor compatibility
The Dionex Aquion IC system supports both chemical and electrolytic suppression technologies. Suppressors remove conductive ions from the eluent to increase analyte signal while decreasing background noise. This enables the highest possible sensitivity and lowest detection limits for conductivity detection.
Optional column heater
Get increased precision and run applications at elevated temperatures with an optional column heater. The column heater preheats the eluent prior to entering the column and maintains the column temperature set by the user. Heater temperature can be set between 30°C and 60°C.
Eluent degasser
Eluent degassing reduces baseline noise, ensuring reproducibility and protection of eluents from contamination and decomposition. An optional built-in vacuum degas provides in-line degassing of eluents, and operation can be automated to sense when degassing is required.
CDS Software
The Dionex Aquion IC system comes with the easy-to-use Thermo Scientific Chromeleon Chromatography Data System (CDS). This software platform provides powerful control and data processing for IC and offers eWorkflows that make all our essential applications easily accessible.
Convenient bundles
Dionex Aquion IC bundles for anion or cation analysis include all hardware, software, and consumables to get started, with optional extended warranties available for peace of mind.
The table below provides an overview of the Dionex Aquion IC system specifications. Not all specifications are listed. For a full list of the specifications, download the Dionex Aquion Ion Chromatography system specifications sheet.
Specification |
Dionex Aquion IC System |
|---|---|
Analytical pump and fluidics |
|
Type |
Serial dual-reciprocating pistons, microprocessor-controlled constant stroke, variable speed |
Construction |
Chemically inert, metal-free PEEK pump heads and flow paths compatible with aqueous eluents of pH 0–14 and reversed-phase solvents |
Pump operating pressure |
0–35 MPa (0–5000 psi) |
Flow rate range |
0.00–5.00 mL/min without changing pump heads |
Flow precision |
<0.1%, typically |
Flow accuracy |
<0.1%, typically |
Pressure ripple |
<1% at 13.8 MPa (2000 psi) and 1.0 mL/min |
Eluent on-off valve |
Standard |
Piston seal wash |
Dual-pump head, wash can be continuous when connected to rinse solution supply |
Pressure alarm limits |
Upper limit 0–35 MPa or 0–5000 psi in one unit (MPa or psi) increments; lower limit can be set up to one unit lower than upper limit |
Vacuum degas |
Yes, optional, automatic control |
Eluent bottles |
Polypropylene, up to 4L volume |
Eluent bottle pressure |
Not required |
Columns supported |
2, 3, 4, and 5 mm i.d.; maximum length 250 mm analytical column with 50 mm guard column |
Injection valve |
|
Type |
6-port, 2 position Rheodyne valve, electrically actuated |
Column heater (optional) |
|
Operating temperature range |
30 to 60 °C (86 to 140 °F); minimum 5 °C above ambient; settable range is equal to working range |
Temperature accuracy |
±0.5 °C at sensor, at 40 °C |
Suppressors and control |
|
Chemical suppression |
2 mm and 4 mm anion and cation, membrane suppression bed types |
Displacement chemical suppression |
2 mm and 4 mm anion and cation, membrane suppression bed types |
Electrolytic suppression |
2 mm and 4 mm anion and cation, membrane suppression bed types available |
Electrolytic suppression with external water mode |
2 mm and 4 mm anion and cation, membrane suppression bed types available |
Current control range |
Thermo Scientific Dionex ERS 500 Electrolytically Regenerated Suppressor: 4 mm, 0–500 mA in 1 mA increments 2 mm, 0–150 mA in 1 mA increments |
Salt converter |
Available in 2 and 4 mm versions |
Carbonic acid removal for anions |
Dionex AERS 500 suppressor with Thermo Scientific Dionex CRD 200 Carbonate Removal Device for hydroxide eluents Dionex AERS 500 suppressor with Dionex CRD 300 device for carbonate eluents |
Non-suppressed chromatography |
Yes, supported |
Suppressor wear parts |
None; peristaltic pump and inline filters not required |
Conductivity detector electronics and flow cell |
|
Type |
Microprocessor-controlled digital signal processor |
Cell drive |
8 kHz square wave |
Detector linearity |
1% up to 1 mS |
Maximum cell pressure |
10 MPa (1500 psi) |
Resolution |
0.00238 nS/cm |
Full-scale output ranges |
Digital signal range 0–15000 μS/cm |
Electronic noise |
±0.1 nS when background conductivity is 0–150 µS/cm |
Filter |
Rise times from 0 to 10 s, user selectable |
Temperature compensation |
Fixed at 1.7% per 1 °C at cell temperature |
Temperature range |
Ambient +7 °C, 30 to 55 °C |
Cell electrodes |
Passivated 316 stainless steel; compatible with MSA |
Cell body |
Chemically inert polymeric material |
Cell volume |
<1 µL |
Heat exchanger |
Inert, tortuous path for low axial dispersion |
Autosampler |
|
Automation using autosampler |
For more information about available autosamplers, visit the IC Autosamplers page |
System software |
|
|
Chromeleon CDS, requires Microsoft® Windows® 10 |
Physical specifications |
|
Dimensions (h × w × d) |
56.1 cm × 22.4 cm × 53.3 cm (22.1 in × 8.8 in × 21 in) |
Weight |
24.5 kg (54 lb) |
Power requirements |
100 to 240 V AC, 50 to 60 Hz, autoranging |
Operating temperature |
4–40 °C (40–104 °F); cold-room-compatible (4 °C) as long as system power remains on |
Operating humidity range |
5–95% relative, noncondensing |
Control modes |
Full control through Chromeleon CDS software; alternative control through TTL or relay closures; two relay outputs, two TTL outputs, four programmable inputs |
USB communication protocol |
One USB input; one built-in two-output USB hub |
Leak detection |
Built-in, optical sensor |
For Research Use Only. Not for use in diagnostic procedures.