On this page:
Increase drug product recovery
Fill-finish is the final step in vaccine or biologic production, where minimizing loss and avoiding contamination are critical. Filling assemblies, consisting of a surge bag, tubing, and filling needles, are delivered into the isolator through a secure port using a beta bag.
Thermo Fisher Scientific offers configurable fill-finish solutions compatible with leading isolator manufacturers, an ideal choice for BioPharma and CDMOs. As a widely used consumables supplier for IMA TILE-X, we offer trusted support to help ensure integrity, excellent lead times, and a range of single-use consumables for compounding, sterile filtration (PUPSIT), and aseptic filling applications.
Our team of specialists can assist with customizable, engineered-to-order solutions to meet your specific needs, helping to ensure high product quality and optimized workflow.
Thermo Scientific fill-finish solutions offer:
  • Increased product recovery: Labtainer Pro BioProcess Container (BPC) design enables optimal drainage with very high product recovery
  • Integrity assurance: 100% assembly helium integrity test with 10 μm detection limit to help minimize leakage risk
  • Improved setup process: BioTitan Retention Device can simplify unpacking steps, compared to traditional cable ties, and helps reduce risk of leaks and connection failures
  • Exceptional lead times: Standard and configurable product designs
  • High-quality film: Rigorously tested single-use Aegis 5-14 film for durability
Thermo Fisher is now the widely used fill-finish supplier for the IMA TILE-X gloveless isolator. The Thermo Scientific Fill Finish Solution offers superior helium integrity testing and exceptional product recovery, enabling high-performance results. Designed for seamless compatibility with IMA TILE-X gloveless technology, it helps deliver excellent reliability and efficiency.
Single-use assemblies for fill-finish processes
The fill-finish product family offers configurable, single-use assemblies from a standard component catalog for drug product final fill applications.
Compounding assemblies
Discover our selection of 2D and 3D bags designed for powders and liquids, with optional integrated filtration. These purpose-built bags and tubing maintain sterility and support maximum product recovery. Each bag is pressure tested with helium gas to help ensure product integrity.
Filtration assembly
Pre-Use Post-Sterilization Integrity Testing (PUPSIT) compliant double filtration assemblies to support final fill-finish. Available to support a variety of volumes across your workflow.
Filling assemblies
Built from high integrity components designed to minimize leak and contamination risk. Filling assemblies are comprised of surge bags, tubing, and filling needles that are aseptically delivered into the isolator through a beta bag.
Experience the fill-finish 3D tour
Explore this interactive tool to discover how our fill-finish solution can significantly enhance drug product recovery and provide exceptional lead times. Our systems are designed to be highly customizable and configurable to meet diverse requirements for optimal efficiency and quality in aseptic filling operations.
Labtainer Pro System
The Labtainer Pro is a single-use bioprocess container system designed for secure fluid handling and storage with minimal product retention. It integrates the BioTitan retention device for enhanced reliability and ease of use.
BioTitan Retention Device
The BioTitan Retention Device helps enhance single-use assemblies with a secure 360-degree seal, replacing traditional cable ties. With zero leaks reported as of 2021, the BioTitan device helps eliminate sharp edges and simplify setup for improve reliability and integrity in bioprocessing workflows.
Beta bag
Available in a variety of bag sizes and transfer port diameters to help securely deliver your fill-finish assembly into the isolator without introducing contamination. Updated packaging is designed to prevent accidental damage and reduce waste.
Frequently asked questions
Fill-finish solutions are used to formulate and deliver sterile drug products like a biologics or vaccines aseptically into vials or prefilled syringes. They consist of bioprocessing containers or bags, surge bags, tubing, and filling needles, and a beta bag. The beta bag delivers the filling assembly into the isolator.
Fill-finish solutions include compounding, filtration, and filling assemblies all designed to maintain an aseptic environment. Compounding takes dry or concentrated API and mixes it with other components to create a stable formulation. To reduce the risk of microbial or residual contamination, filtration may be used as the next step in the fill-finish process, where filling assemblies—including the drug surge bag, tubing assemblies, and filling needles—are delivered into a filling isolator through a beta bag.
Aseptic filling is the process of transferring sterile products into sterile containers in a controlled environment to prevent contamination. The process includes sterilizing the product, containers, and equipment, followed by filling and sealing the containers using specialized machinery within an aseptic environment, often maintained by isolators or RABS. Single-use fill solutions like bags reduce risk of contamination and speed equipment turnover.
We have a large network of global manufacturing facilities that ensure redundancy in manufacturing and faster delivery times. Many of our fill-finish products are made-to-order using our Global Standard Component Library (GSCL), which includes pre-qualified components with standard claims. The GSCL features our most widely used components, selected for their broad applicability and universal relevance, and stocked within our network for efficient production and delivery.
Bioprocessing films
Bioprocessing films are critical in the bioprocessing workflow, providing sterile, durable containment from cell cultivation and media storage to fill finish. We offer high-quality, single-use films that help ensure product integrity, reduce contamination risk, and enhance efficiency.
Sterilizing Grade Filters
Integrate LifeASSURE sterilizing grade filters into your Thermo Scientific single-use assemblies—offering you the confidence of quality filtration performance, improved workflow integration, and reliable global supply.
Explore BioProcess Insights
The Bioprocess Insights Hub by Thermo Fisher Scientific is designed for biomanufacturing professionals, offering educational resources to optimize single-use bioprocessing workflows. It features articles, guides, and case studies on topics such as assembly design, contamination mitigation, and technology transfer. Resources include application notes, brochures, white papers, videos, and webinars, covering areas like process optimization, scalability, quality, and sustainability.
Speak with a technical specialist about your process needs
For research use or further manufacturing. Not for diagnostic use or direct administration into humans or animals.