HT Supplement
HT Supplement liquid is a mixture of sodium hypoxanthine and thymidine. After dilution, HT Supplement is suitable for use as a rescue medium. The supplement provides preformed purines and pyrimidines to overcome the effects of residual intracellular aminopterin. Once the
de novo biosynthesis pathway for nucleosides has been reestablished, HT supplementation can be discontinued.