Dulbecco's phosphate-buffered saline (DPBS) can be used to provide a buffer system to maintain cell culture media in the physiological pH range, or to irrigate, transport, or dilute fluid while maintaining cell tonicity and viability. Gibco DPBS is manufactured in our state-of-the-art cGMP, ISO-certified facilities, so you can always count on the highest quality and consistency.
Gibco DPBS is available in multiple formulations and formats to meet your specific research needs, including calcium- and magnesium-free versions for washing cells prior to trypsinization.
We also have formulations to enable you to prepare for the clinic! Be sure to check out CTS DPBS (with calcium chloride and magnesium chloride) and CTS DPBS (w/o calcium chloride and magnesium chloride)