Fast, simple, and trusted workflow for your Sanger sequencing applications
Our simple and fast Sanger sequencing workflow can be completed in less than one workday, from sample to answer. We provide complete workflow solutions with an instrument that uses our gold-standard technology, reagents, consumables, analysis software, and world-class technical support to address the ongoing progression of scientific questions, leading to answers and solutions.
Learn about the Sanger method for DNA sequencing, Sanger sequencing chemistries, and capillary electrophoresis technology.
Save time with the Invitrogen™ Primer Designer™ Tool by selecting from ~650,000 primer pairs targeting the human exome and human mitochondrion genome.
Discover primary and secondary data analysis tools and software solutions for interpreting Sanger sequencing results.
Isolate DNA
Isolation of DNA from samples of interest, a critical first step for any Sanger sequencing application.
Perform PCR
Amplify the DNA region of interest: select from predesigned primer pairs or design your own.
Generate products
Generate sequencing products by cycle sequencing; purify reactions to remove fluorescent ddNTP.
Run sequence
The products of the sequencing reaction are injected into capillaries filled with polymer to perform capillary electrophoresis.
Analyze data
Translate raw data into corresponding electropherograms using downstream software applications.
Resources
Interested in commercializing your assay?
We can help ›
Support
New Capillary Electrophoresis Instruments Support Center
Find tips, troubleshooting help, and resources for support related to your capillary electrophoresis instruments.