Search
Multinucleate HeLa cell in metaphase: DAPI, anti–bovine α-tubulin, mouse monoclonal 236-10501, biotin-XX conjugate and Alexa Fluor® 568 streptavidin. A multinucleate HeLa cell in metaphase that was fixed and then stained with a combination of fluorescent dyes. The chromosomes were stained with DAPI (Cat. no. D1306, D3571, D21490). The cytoskeleton was detected with the biotin-XX conjugate of mouse monoclonal anti–α-tubulin antibody (Cat. no. A21371), which was then visualized with red-fluorescent Alexa Fluor® 568 streptavidin (Cat. no. S11226). The multiple-exposure image was acquired using filter sets appropriate for rhodamine and DAPI.
1% Agarose gel containing 16S and 23S ribosomal RNA (rRNA). SYBR® Green II RNA gel stain. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
Increase in accuracy and precision using the Qubit® Go ›
Performance of the Qubit® dsDNA HS Assay Go ›
Endothelial cells. Go ›
Mouse Anti-Alpha Tubulin Monoclonal Antibody (Cat. No. A11126) Go ›
Muntjac fibroblast labeled with probes for actin and the nucleus. Go ›
Multicolor fluorescence analysis of muntjac fibroblasts. Go ›
A prometaphase muntjac skin fibroblast stained with Alexa Fluor® 350 phalloidin, an anti–a-tubulin antibody and an anti–cdc6 peptide antibody. Go ›
Simultaneous detection of expression of five genes in a whole mount Drosophila embryo by fluorescence in situ hybridization (FISH) with five RNA probes Go ›