Informational:Inquire about OEM or Commercial Supply version of this product here.
Thermo Scientific™

Micrococcal Nuclease (300 U/μL)

Catalog number: EN0181
Have Questions?
Thermo Scientific™

Micrococcal Nuclease (300 U/μL)

Catalog number: EN0181
Have Questions?
Catalog Number
EN0181
Unit Size
8,000 units
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
EN01818,000 units
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Micrococcal Nuclease (S7 Nuclease) is an endo-exonuclease that digests single-stranded and double-stranded DNA and RNA.
Micrococcal Nuclease (S7 Nuclease) is a relatively nonspecific endo-exonuclease that digests single-stranded and double-stranded nucleic acids, but is more active on single-stranded substrates. Cleavage of DNA or RNA occurs preferentially at AT or AU-rich regions, yielding mononucleotides and oligonucleotides with terminal 3'-phosphates. The enzyme's activity is strictly dependent on Ca2+.
Applications

• Hydrolysis of nucleic acids in crude cell-free extracts
• Sequencing of RNA (see​ Reference 2)
• Studies of chromatin structure
• A model for protein folding and for structure-function studies
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Concentration
300 U/μL
Enzyme
Nuclease
Quantity
8000 U
Product Type
Micrococcal Nuclease

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text