Informational:Inquire about OEM or Commercial Supply version of this product here.
Thermo Scientific™
RNase A/T1 Mix
Catalog number: EN0551
Have Questions?
Thermo Scientific™
RNase A/T1 Mix
Catalog number: EN0551
Have Questions?
Catalog Number
EN0551
Unit Size
1 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
EN05511 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific RNase A/T1 Mix combines the RNA degradation activity of both RNase A and RNase T1. The RNase A specifically hydrolyzes RNA at C and U residues; RNase T1 specifically hydrolyzes RNA at G residues.
Highlights
• Higher level of RNA degradation than with RNase A and RNase T1 separately
• Free of DNase activity - It is not necessary to heat the mix before use
Applications
• Removal of RNA from DNA preparations
• Removal of RNA from recombinant protein preparations
• Ribonuclease protection assays
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Enzyme
RNase
Product Type
RNase A/T1 Mix