Search Thermo Fisher Scientific
Leverage Applied Biosystems microarray technology to develop microarray products for applications in diagnostics, forensics, animal testing, industrial testing, food testing, and more. We have made this breakthrough technology available to companies like yours, so you can create tailored products and services for your unique markets and customers, and realize unprecedented opportunity.
This flexible technology can be tailored to fit your needs. One platform can enable:
GeneChip Scanner 3000 | GeneTitan Multi-Channel (MC) Instrument | |
![]() | ![]() | |
Regulatory status | Available as RUO or IVD in most regions | RUO only |
Key applications | Gene expression profiling Cytogenetic analysis | Genotyping |
Benefits | Enables higher-density arrays | Integrates hybridization, washing, and imaging into a single instrument |
The Axiom Precision Medicine Diversity Array (PMD Array) is a comprehensive imputation-aware genotyping array, which provides valuable scientific insights into human disease with GWAS markers and relevant variants. It offers >850,000 markers selected for high genomic coverage from the 1000Genomes Project Phase III.
Obtain a gene-level view of the human transcriptome with the Clariom S Assays for human samples. Clariom S Assays serve as a next generation transcriptome-wide gene-level expression profiling tool, which allows for a fast, simple, and scalable path to generating the results you need for your research.
Available for human, mouse, and rat, Clariom D Assays allow to generate high-fidelity biomarker signatures quickly and easily with a design that provides intricate transcriptome‑wide, gene- and exon-level expression profiles, including the ability to detect alternative splicing events of coding and long noncoding RNA, in a single three-day experiment.
For Research Use Only. Not for use in diagnostic procedures.