Search
Primary XPS region: Cr2p
Overlapping regions: Te3d, Zn LMM
Binding energies of common chemical states:
| Chemical state | Binding energy Cr2p3/2 |
|---|---|
| Cr metal | 574.3 eV |
| Cr(III) oxide | ~576 eV |
| Cr(VI) oxide | ~580 eV |
Experimental information
Interpretation of XPS spectra
References
About this element
Symbol: Cr
Date of discovery: 1797
Name origin: Greek chrôma
Appearance: gray
Discoverer: Louis Vauquelin
Obtained from: chromite
Melting point: 2180 K
Boiling point: 2944 K
Density[kg/m3]: 7140
Molar volume: 7.23 × 10-6 m3/mol
Protons/Electrons: 24
Neutrons: 28
Shell structure: 2,8,13,1
Electron configuration: [Ar]3d54s1
Oxidation state: 6,4,3,2
Crystal structure: cubic
Chromium is a malleable hard metal best known for its attractive luster when polished. Chromium gets its name from the Greek word for color due to the many colorful compounds that can be made from it. During the 1800s, chromium was used mainly as a component in paints and tanning salts for leather. Today, 85% of its use is in metallic alloys, such as stainless steel and anodized aluminum. Traces of chromium can be found in gemstones, such as ruby and emerald. Chromium is a common element and is used in producing synthetic gems.
최적의 시스템 성능을 보장하기 위해 세계 수준의 현장 서비스 전문가 네트워크, 기술 지원 및 인증된 예비 부품을 제공해드립니다.