Nickel • Transition Metal
Primary XPS region: Ni2p
Overlapping regions: Fe LMM
Binding energies of common chemical states:
Chemical stateBinding energy Ni2p3/2
Ni metal852.6 eV
NiO853.7 eV
Ni(OH)2855.6 eV
Experimental information
  • Nickel oxide is readily reduced by Ar+ sputtering.
    • Use lowest possible ion beam energy when attempting to clean oxide sample surface or during depth profiling.
      • Minimizes (but does not completely prevent) reduction effect.
Interpretation of XPS spectra
  • Ni2p peak has significantly split spin-orbit components (Δmetal=17.3eV).
  • Ni metal spectrum has complex shape.
    • Mixture of core level and satellite features.
    • Satellite features not to be confused with oxidized nickel peaks.
High Resolution Ni2p Spectrum Ni Metal
  • Ni compounds can also have complex, multiplet-split peaks.
Ni2p Spectra Ni Oxide Hydroxide
Nickel • Transition Metal
Element Crystal Fcc
About this element
Symbol: Ni
Date of discovery: 1751
Name origin: German kupfernickel
Appearance: white
Discoverer: Axel Fredrik Cronstedt
Obtained from: pentlandite
Melting point: 1728 K
Boiling point: 3186 K
Density[kg/m3]: 8908
Molar volume: 6.59 × 10-6 m3/mol
Protons/Electrons: 28
Neutrons: 31
Shell structure: 2,8,16,2
Electron configuration: [Ar]3d84s2
Oxidation state: 2,3
Crystal structure: cubic face centered
The consumption of nickel can be traced back to 3500 BC. The majority of the supply of nickel is believed to be located in the Earth’s core, while Canada, Russia, New Caledonia, Australia, Cuba, and Indonesia house accessible deposits of nickel. Most of the nickel consumed in the Western World is used to make austenitic stainless steel. Nickel steel is used for armor plates and vaults. Nickel is also used in the five-cent coins in the United States and Canada (called nickels). It also makes up crucibles that are used in chemical laboratories. One of its isotopes, nickel-56, is produced in type II supernova. The light curve of the supernova is associated with the decay of nickel-56 to cobalt-56 and then to iron-56. Nickel sulfide fume and dust is believed to be carcinogenic and nickel carbonyl gas is extremely toxic.
Products
Nexsa G2 XPS
  • Micro-focus X-ray sources
  • Unique multi-technique options
  • Dual-mode ion source for monoatomic & cluster ion depth profiling
Style Sheet for Instrument Cards Original
ESCALAB QXi XPS
  • High spectral resolution
  • Multi-technique surface analysis
  • Extensive sample preparation and expansion options
Contact us
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
Style Sheet to change H3 to p with em-h3-header class
Style Sheet to change H2 style to p with em-h2-header class
Style Sheet for Products Table Specifications
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
Style Sheet to change H3 to p with em-h3-header class
Style Sheet to change H2 style to p with em-h2-header class
Style Sheet for Products Table Specifications
Fix for left nav
Style Sheet for Support and Service footer
Style Sheet for Fonts
Style Sheet for Cards
Electron microscopy services for
the materials science
To ensure optimal system performance, we provide you access to a world-class network of field service experts, technical support, and certified spare parts.