As semiconductor device design approaches smaller nodes, it becomes more and more difficult to ramp manufacturing yields. These smaller nodes, as well as more complex 3D architectures, result in added steps and more complexity in the design process. Hundreds of steps are involved, and any defect or electrical fault that occurs in any one step will slow down the total production time. Since time-to-market and time-to-yield are both crucial for the commercial success of any new semiconductor design, metrology and inspection tools are needed to make sure each of these steps is optimized. This ensures the maximum yield can be guaranteed and maintained.
Yield analysis must be carried out as quickly and as inexpensively as possible. However, given the many steps involved in device manufacture, this can often be an extremely challenging proposition. For example, in an advanced logic device, many different measurements need to be taken, such as the finFET gate height, fin height, and sidewall angle. Performing these measurements in a cost-effective and timely manner requires advanced characterization tools designed specifically for semiconductor analysis. By using automated SEM or (S)TEM based metrology instruments, manufacturers can provide accurate and timely semiconductor inspection, lower manufacturing costs, and shorten the product development cycle.
Thermo Fisher Scientific offers a suite of next-generation products with advanced analytical capabilities for semiconductor metrology and inspection. These solutions are designed to help increase productivity in semiconductor fabrication labs by improving quality control and yield in the manufacture of logic, 3D NAND, DRAM, analog, power and display devices.
Semiconductor metrology & yield ramp workflow examples
Style Sheet for Komodo Tabs
Style Sheet to change H2 style to p with em-h2-header class
Techniques
Aislamiento de fallo térmico
La distribución desigual de la disipación de energía local puede causar aumentos de temperatura importantes y localizados, lo que provoca un fallo del dispositivo. Ofrecemos soluciones únicas para el aislamiento de fallos térmicos con termografía infrarroja de alta sensibilidad (LIT).
Más información ›
Metrología de TEM
Las rutinas de metrología TEM avanzadas y automatizadas ofrecen una precisión significativamente mayor que los métodos manuales. Esto permite a los usuarios generar grandes cantidades de datos estadísticamente relevantes, con una especificidad de nivel subangstrom, sin fallos del operador.
Más información ›
Preparación de muestras de dispositivos semiconductores
Los sistemas DualBeam de Thermo Scientific proporcionan una preparación precisa de las muestras de TEM para el análisis a escala atómica de dispositivos semiconductores. La automatización y las tecnologías de aprendizaje automático avanzado producen muestras de alta calidad, en la ubicación correcta y con un bajo costo por muestra.
Más información ›
Metrología de SEM
La microscopía electrónica de barrido proporciona datos de metrología precisos y fiables a escala nanométrica. La metrología SEM automatizada de resolución ultraalta permite un tiempo de producción y un de comercialización más rápidos para aplicaciones de memoria, lógica y almacenamiento de datos.
Más información ›
Adquisición de imágenes y análisis de semiconductores
Thermo Fisher Scientific ofrece microscopios electrónicos de barrido para todas las funciones de un laboratorio de semiconductores, desde tareas generales de adquisición de imágenes hasta técnicas avanzadas de análisis de fallos que requieren mediciones precisas de contraste de tensión.
Más información ›
Reestructuración de dispositivo
La contracción del tamaño de las características, junto con los resultados de diseño y arquitectura avanzados provocan fallos cada vez más complicados para los semiconductores. La reestructuración sin daños de los dispositivos es una técnica crucial para la detección de errores y fallos eléctricos interiores.
Más información ›
Aislamiento de fallo térmico
La distribución desigual de la disipación de energía local puede causar aumentos de temperatura importantes y localizados, lo que provoca un fallo del dispositivo. Ofrecemos soluciones únicas para el aislamiento de fallos térmicos con termografía infrarroja de alta sensibilidad (LIT).
Más información ›
Metrología de TEM
Las rutinas de metrología TEM avanzadas y automatizadas ofrecen una precisión significativamente mayor que los métodos manuales. Esto permite a los usuarios generar grandes cantidades de datos estadísticamente relevantes, con una especificidad de nivel subangstrom, sin fallos del operador.
Más información ›
Preparación de muestras de dispositivos semiconductores
Los sistemas DualBeam de Thermo Scientific proporcionan una preparación precisa de las muestras de TEM para el análisis a escala atómica de dispositivos semiconductores. La automatización y las tecnologías de aprendizaje automático avanzado producen muestras de alta calidad, en la ubicación correcta y con un bajo costo por muestra.
Más información ›
Metrología de SEM
La microscopía electrónica de barrido proporciona datos de metrología precisos y fiables a escala nanométrica. La metrología SEM automatizada de resolución ultraalta permite un tiempo de producción y un de comercialización más rápidos para aplicaciones de memoria, lógica y almacenamiento de datos.
Más información ›
Adquisición de imágenes y análisis de semiconductores
Thermo Fisher Scientific ofrece microscopios electrónicos de barrido para todas las funciones de un laboratorio de semiconductores, desde tareas generales de adquisición de imágenes hasta técnicas avanzadas de análisis de fallos que requieren mediciones precisas de contraste de tensión.
Más información ›
Reestructuración de dispositivo
La contracción del tamaño de las características, junto con los resultados de diseño y arquitectura avanzados provocan fallos cada vez más complicados para los semiconductores. La reestructuración sin daños de los dispositivos es una técnica crucial para la detección de errores y fallos eléctricos interiores.
Más información ›
Samples
Materiales semiconductores y caracterización de dispositivos
A medida que los dispositivos semiconductores se reducen y se vuelven más complejos, se necesitan nuevos diseños y estructuras. Los flujos de trabajo de análisis en 3D de alta productividad pueden reducir el tiempo de desarrollo de dispositivos, maximizar el rendimiento y garantizar que los dispositivos satisfacen las necesidades futuras del sector.
Más información ›
Products
Hoja de estilo para tarjetas originales instrumentos
Helios 5 PFIB DualBeam
  • Preparación de muestras STEM y TEM sin galio
  • Subsuperficie multimodal e información 3D
  • Columna FIB con plasma xenón de 2,5 μA de última generación
Helios 5 DualBeam
  • Preparación de muestras de TEM ultrafina, de alta calidad y completamente automatizada
  • Subsuperficie de alto rendimiento, alta resolución y caracterización en 3D
  • Capacidades de rápida creación de prototipos a nanoescala
Metrios AX TEM
  • Opciones de automatización compatibles con calidad, uniformidad, metrología y OPEX reducido
  • Aprovecha el aprendizaje automático para excelentes funciones automáticas y reconocimiento de funciones
  • Flujos de trabajo para preparación de laminillas in-situ y ex-situ
Scios 2 DualBeam
  • Soporte completo de muestras magnéticas y no conductoras
  • Subsuperficie de alto rendimiento y caracterización en 3D
  • Facilidad de uso y capacidades de automatización avanzadas
ExSolve WTP DualBeam
  • Puede preparar laminillas específicas de sitios de 20 nm de grosor en obleas completas de hasta 300 mm de diámetro
  • Soluciona las necesidad de muestreo automatizado de alto rendimiento en nodos de tecnología avanzada
Verios 5 XHR SEM
  • SEM monocroma para resolución de subnanómetros sobre el rango de energía completo de 1 keV a 30 keV
  • Fácil acceso a la energía de recepción de haz, a un nivel tan bajo como 20 eV
  • Excelente estabilidad con fase piezoeléctrica como estándar
Quattro ESEM
  • Tecnología de obtención de imágenes por microscopía electrónica de barrido (SEM) con cañón de emisión de campo (FEG) de alta resolución con versatilidad absoluta y capacidad medioambiental única (ESEM)
  • Consulte la información completa de todas las muestras con obtención de imágenes SE y BSE en cada modo de funcionamiento
Apreo 2 SEM
  • SEM de alto rendimiento para resolución de subnanómetro o nanómetro completo
  • Detector de retrodispersión de T1 incorporado en la columna para contraste de materiales sensibles de velocidad TV
  • Excelente rendimiento en distancias de trabajo largas (10 mm)
SEM de escritorio Phenom ProX
  • SEM de escritorio de alto rendimiento con detector EDS integrado
  • Resolución de <8 nm (SE) y <10 nm (BSE); aumento de hasta 150.000x
  • Detector SE opcional
Sistema ELITE
  • Completamente no destructivo
  • Identifica rápidamente el componente defectuoso en la tarjeta de montaje y obtener la disposición exacta
  • Localiza el defecto en x-y con la precisión del micrómetro, con una ubicación de profundidad exacta de 20 µm
AutoTEM 5
  • Preparación de muestras S/TEM in situ totalmente automatizada
  • Compatible con geometría invertida, plana y "top-down" (arriba-abajo)
  • Flujo de trabajo altamente configurable
  • Interfaz de usuario intuitiva, fácil de utilizar
Software Auto Slice and View 4.0
  • Cortes en secciones en serie automatizadas para DualBeam
  • Adquisición de datos multimodales (SEM, EDS, EBSD)
  • Capacidades de edición sobre la marcha
  • Ubicación del corte en los bordes
Contact us