    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • CD66b (CEACAM8) Antibodies
          The product (cat # {{productObj.productId}}) that you're looking for has been discontinued and replaced with {{productObj.replacementProductId}}. The product (cat # {{productObj.productId}}) that you're looking for has been discontinued. Please contact Technical Support with any questions.
          Invitrogen
          CD66b Monoclonal Antibody (G10F5), PE, eBioscience™
          1 Published Figure
          8 References
          View all (18) CD66b (CEACAM8) antibodies
          Tech Support
          Cite CD66b Monoclonal Antibody (G10F5), PE, eBioscience™
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          • Published Figures (1)
          CD66b Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD66b Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          CD66b Antibody (12-0666-41) in Flow
          Staining of normal human peripheral blood cells with Mouse IgM Isotype Control PE (Product # 12-4752) (blue histogram) or Anti-Human CD66b PE (purple histogram). Cells in the granulocyte gate were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          CD66b Antibody in Flow Cytometry (Flow)
          CD66b Antibody in Flow Cytometry (Flow)
          Product Details
          12-0666-41
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          -
          View 2 publications 2 publications
          Flow Cytometry (Flow)
          5 µL (0.125 µg)/test
          View 6 publications 6 publications
          Product Specifications
          Published species
          Human, Mouse
          Host/Isotype
          Mouse / IgM, kappa
          Class
          Monoclonal
          Type
          Antibody
          Clone
          G10F5
          Conjugate
          PE PE PE
          • APC 
          • FITC
          • PE-Cyanine7
          • Request custom conjugation
          Excitation/Emission Max
          565/576 nm View spectra spectra
          Form
          Liquid
          Concentration
          5 µL/Test
          Storage conditions
          4°C, store in dark, DO NOT FREEZE!
          Shipping conditions
          Wet ice
          RRID
          AB_2572566
          Product Specific Information
          Description: This G10F5 monoclonal antibody reacts with human CD66b, one of four members of the CD66 family of granulocyte-specific activation antigens. Expressed on neutrophils and eosinophils, CD66b is upregulated following stimulation. This glycosyl-phosphatidylinositol-linked cell surface receptor plays a role in regulating neutrophil activation and adhesion to fibronectin, galectin-3, and E-selectin.
          Applications Reported: This G10F5 antibody has been reported for use in flow cytometric analysis.
          Applications Tested: This G10F5 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 µL (0.125 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
          Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.
          Filtration: 0.2 µm post-manufacturing filtered.
          Target Information
          This gene encodes a member of the carcinoembryonic antigen (CEA) gene family, which belongs to the immunoglobulin superfamily. Two subgroups of the CEA family, the CEA cell adhesion molecules and the pregnancy-specific glycoproteins, are located within a 1.2 Mb cluster on the long arm of chromosome 19. Eleven pseudogenes of the CEA cell adhesion molecule subgroup are also found in the cluster. The encoded protein was originally described in bile ducts of liver as biliary glycoprotein. Subsequently, it was found to be a cell-cell adhesion molecule detected on leukocytes, epithelia, and endothelia. The encoded protein mediates cell adhesion via homophilic as well as heterophilic binding to other proteins of the subgroup. Multiple cellular activities have been attributed to the encoded protein, including roles in the differentiation and arrangement of tissue three-dimensional structure, angiogenesis, apoptosis, tumor suppression, metastasis, and the modulation of innate and adaptive immune responses. Multiple transcript variants encoding different isoforms have been reported, but the full-length nature of all variants has not been defined.
          How to use the Panel Builder
          Argentina flag icon
          Argentina
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline