    • See all product categories
  • Applications
    • Cell Analysis
    • Protein Biology
    • Lab Informatics
    • 360° CDMO and CRO Solutions
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
          • Primary Antibodies ›
          • CD8b Antibodies
          Invitrogen
          CD8b Monoclonal Antibody (eBioH35-17.2 (H35-17.2)), eBioscience™
          8 Published Figures
          27 References
          View all (76) CD8b antibodies
          Tech Support
          Cite CD8b Monoclonal Antibody (eBioH35-17.2 (H35-17.2)), eBioscience™
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          • Published Figures (8)
          CD8b Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD8b Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          FIGURE: 1 / 9
          CD8b Antibody (14-0083-85) in Flow
          Staining of BALB/c splenocytes with 0.25 µg of Rat IgG2b Isotype Control Purified (Product # 14-4031-82) (open histogram) or 0.25 µg of Purified Anti-Mouse CD8b Purified (filled histogram) followed by Anti-Rat IgG FITC (Product # 11-4811-85). Cells in the lymphocyte gate were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          CD8b Antibody in Flow Cytometry (Flow)
          CD8b Antibody in Flow Cytometry (Flow)
          CD8b Antibody in Flow Cytometry (Flow)
          CD8b Antibody in Flow Cytometry (Flow)
          CD8b Antibody in Flow Cytometry (Flow)
          CD8b Antibody in Flow Cytometry (Flow)
          CD8b Antibody in Flow Cytometry (Flow)
          CD8b Antibody in Flow Cytometry (Flow)
          CD8b Antibody in Flow Cytometry (Flow)
          Product Details
          14-0083-85
          Applications
          Tested Dilution
          Publications
          Immunohistochemistry (IHC)
          -
          View 6 publications 6 publications
          Immunohistochemistry (Frozen) (IHC (F))
          Assay-Dependent
          View 3 publications 3 publications
          Flow Cytometry (Flow)
          0.5 µg/test
          View 17 publications 17 publications
          Immunoprecipitation (IP)
          Assay-Dependent
          -
          Inhibition Assays (Inhibition)
          -
          View 1 publication 1 publication
          Product Specifications
          Species Reactivity
          Mouse
          Published species
          Mouse
          Host/Isotype
          Rat / IgG2b, kappa
          Class
          Monoclonal
          Type
          Antibody
          Clone
          eBioH35-17.2 (H35-17.2)
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • Alexa Fluor 488 
          • APC 
          • APC-eFluor 780
          • eFluor 450
          • FITC
          • PE 
          • PE-Cyanine5
          • PE-Cyanine7
          • PerCP-eFluor 710
          • Super Bright 436
          • Super Bright 600
          • Super Bright 702
          • Request custom conjugation
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          500 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.2
          Contains
          0.09% sodium azide
          Storage conditions
          4°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_657759
          Product Specific Information
          Description: The eBioH35-17.2 monoclonal antibody reacts with the mouse CD8 beta molecule. The CD8 beta chain associates with the CD8 alpha chain to form the CD8 alpha/beta heterodimer expressed on the surface of a majority of thymocytes, and on peripheral cytotoxic alpha beta TCR T cells. CD8 binds to MHC class I and plays a role in T cell development and activation of mature T cells.
          Applications Reported: This eBioH35-17.2 (H35-17.2) antibody has been reported for use in flow cytometric analysis, immunoprecipitation, and immunohistochemical staining of frozen tissue sections.
          Applications Tested: This eBioH35-17.2 (H35-17.2) antibody has been tested by flow cytometric analysis of mouse splenocytes and thymocytes. This can be used at less than or equal to 0.5 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
          Purity: Greater than 90%, as determined by SDS-PAGE.
          Aggregation: Less than 10%, as determined by HPLC.
          Filtration: 0.2 µm post-manufacturing filtered.
          Target Information
          The CD8B antigen is a cell surface glycoprotein found on most cytotoxic T lymphocytes that mediates efficient cell-cell interactions within the immune system. The CD8 antigen, acting as a coreceptor, and the T-cell receptor on the T lymphocyte recognize antigens displayed by an antigen presenting cell (APC) in the context of class I MHC molecules. The functional coreceptor is either a homodimer composed of two alpha chains, or a heterodimer composed of one alpha and one beta chain. Both alpha and beta chains share significant homology to immunoglobulin variable light chains. This gene encodes the CD8 beta chain isoforms. Multiple alternatively spliced transcript variants encoding distinct membrane associated or secreted isoforms have been described. A pseudogene, also located on chromosome 2, has been identified.
          Bioinformatics
          Protein Aliases: CD8; CD8a; CD8alpha; CD8b; CD8beta; Leu-2; Lymphocyte antigen 3; MAL; p32; T-cell membrane glycoprotein Ly-3; T-cell surface glycoprotein CD8 beta chain; T-cell surface glycoprotein Lyt-3
          View more View less
          Gene Aliases: Cd8b; Cd8b1; Ly-3; Ly-C; Lyt-3; Lyt3
          View more View less
          UniProt ID: (Mouse) P10300
          View more View less
          Entrez Gene ID: (Mouse) 12526
          View more View less
          Function(s)
          protein binding defense/immunity protein immunoglobulin receptor superfamily
          Process(es)
          immune system process
          • Trademarks
          • Fair Trade Fair Trade
          Korea flag icon
          Korea
          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat
          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat
          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat
          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910
          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783
          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)
          ISMS Logo
          Your items have has been added!
          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline