    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CD166 (ALCAM) Antibodies
          Invitrogen
          CD166 (ALCAM) Monoclonal Antibody (eBioALC48), eBioscience™
          18 Published Figures
          8 References
          View all (73) CD166 (ALCAM) antibodies
          Tech Support
          Cite CD166 (ALCAM) Monoclonal Antibody (eBioALC48), eBioscience™
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          • Published Figures (18)
          CD166 (ALCAM) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD166 (ALCAM) Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          FIGURE: 1 / 19
          CD166 (ALCAM) Antibody (14-1661-81) in Flow
          Staining of C57BL/6 splenocytes with 0.125 µg of Rat IgG2a kappa Isotype Control Purified (Product # 14-4321-82) (open histogram) or 0.125 µg of Anti-Mouse CD166 (ALCAM) Purified (filled histogram) followed by Anti-Rat IgG Biotin (Product # 13-4813-85) and Streptavidin PE (Product # 12-4317-87). Cells in the lymphocyte gate were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          CD166 (ALCAM) Antibody in Flow Cytometry (Flow)
          CD166 (ALCAM) Antibody in Immunohistochemistry (IHC)
          CD166 (ALCAM) Antibody in Immunohistochemistry (IHC)
          CD166 (ALCAM) Antibody in Immunohistochemistry (IHC)
          CD166 (ALCAM) Antibody in Immunohistochemistry (IHC)
          CD166 (ALCAM) Antibody in Immunohistochemistry (IHC)
          CD166 (ALCAM) Antibody in Immunohistochemistry (IHC)
          CD166 (ALCAM) Antibody in Immunohistochemistry (IHC)
          CD166 (ALCAM) Antibody in Immunohistochemistry (IHC)
          CD166 (ALCAM) Antibody in Immunohistochemistry (IHC)
          CD166 (ALCAM) Antibody in Immunohistochemistry (IHC)
          CD166 (ALCAM) Antibody in Immunohistochemistry (IHC)
          CD166 (ALCAM) Antibody in Immunohistochemistry (IHC)
          CD166 (ALCAM) Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          CD166 (ALCAM) Antibody in Flow Cytometry (Flow)
          CD166 (ALCAM) Antibody in Flow Cytometry (Flow)
          CD166 (ALCAM) Antibody in Flow Cytometry (Flow)
          CD166 (ALCAM) Antibody in Flow Cytometry (Flow)
          CD166 (ALCAM) Antibody in Flow Cytometry (Flow)
          Product Details
          14-1661-81
          Applications
          Tested Dilution
          Publications
          Immunohistochemistry (IHC)
          -
          View 2 publications 2 publications
          Immunohistochemistry (Paraffin) (IHC (P))
          Assay-Dependent
          -
          Immunohistochemistry (PFA fixed) (IHC (PFA))
          -
          View 1 publication 1 publication
          Flow Cytometry (Flow)
          0.25 µg/test
          View 5 publications 5 publications
          Product Specifications
          Species Reactivity
          Mouse
          Published species
          Mouse
          Host/Isotype
          Rat / IgG2a, kappa
          Class
          Monoclonal
          Type
          Antibody
          Clone
          eBioALC48
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • APC 
          • FITC
          • PE 
          • Request custom conjugation
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          50 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.2
          Contains
          0.09% sodium azide
          Storage conditions
          4°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_823128
          Product Specific Information
          Description: Activated leukocyte cell adhesion molecule (ALCAM; CD166) is a 105 kD cell-surface adhesion molecule belonging to the Ig superfamily of receptors. ALCAM is a type I transmembrane glycoprotein, possessing five extracellular Ig domains, a transmembrane domain, and a short cytoplasmic tail. ALCAM is involved in homotypic adhesion, as well as heterotypic adhesion with its natural ligand, CD6. A soluble isoform of ALCAM has been shown to inhibit homophilic adhesive interactions. ALCAM is expressed on a variety of cell types, including activated T and B cells, monocyte-derived dendritic cells, and thymic epithelial cells. ALCAM expression has also been used as a marker for the isolation of pluripotent stem cells.
          Applications Reported: This eBioALC-48 antibody has been reported for use in flow cytometric analysis and immunohistochemistry on paraffin-embedded tissue sections.
          Applications Tested: The eBioALC-48 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
          Purity: Greater than 90%, as determined by SDS-PAGE.
          Aggregation: Less than 10%, as determined by HPLC.
          Filtration: 0.2 µm post-manufacturing filtered.
          Target Information
          This gene encodes activated leukocyte cell adhesion molecule (ALCAM), also known as CD166 (cluster of differentiation 166), which is a member of a subfamily of immunoglobulin receptors with five immunoglobulin-like domains (VVC2C2C2) in the extracellular domain. This protein binds to T-cell differentiation antigene CD6, and is implicated in the processes of cell adhesion and migration. Multiple alternatively spliced transcript variants encoding different isoforms have been found.
          Bioinformatics
          Protein Aliases: Activated leukocyte cell adhesion molecule; BEN; CD166; CD166 antigen; FLJ38514; MGC71733; Protein DM-GRASP
          View more View less
          Gene Aliases: AI853494; Alcam; BEN; CD166; DM-GRASP; MuSC; SC1
          View more View less
          UniProt ID: (Mouse) Q61490
          View more View less
          Entrez Gene ID: (Mouse) 11658
          View more View less
          Function(s)
          protein binding
          Process(es)
          cell adhesion axon guidance motor neuron axon guidance
          France flag icon
          France
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline