    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • FOXJ1 Antibodies
          Invitrogen
          FOXJ1 Monoclonal Antibody (2A5), eBioscience™
          Advanced Verification
          74 Published Figures
          123 References
          View all (17) FOXJ1 antibodies
          Tech Support
          Cite FOXJ1 Monoclonal Antibody (2A5), eBioscience™
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          • Published Figures (74)
          • Advanced Verification (1)
          FOXJ1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          FOXJ1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          FIGURE: 1 / 77
          FOXJ1 Antibody (14-9965-80) in IHC (P)
          Immunohistochemistry of paraffin-embedded mouse lung tissue with Anti-Human/Mouse FOXJ1 Purified followed by Anti-Mouse IgG Biotin (Product # 13-4013-85) and Avidin HRP (Product # 18-4100-51) (right).Data provided courtesy of Steven L.Brody. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          FOXJ1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FOXJ1 Antibody in Western Blot (WB)
          FOXJ1 Antibody in Immunocytochemistry (ICC/IF)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunocytochemistry (ICC/IF)
          FOXJ1 Antibody in Immunocytochemistry (ICC/IF)
          FOXJ1 Antibody in Immunocytochemistry (ICC/IF)
          FOXJ1 Antibody in Immunocytochemistry (ICC/IF)
          FOXJ1 Antibody in Immunocytochemistry (ICC/IF)
          FOXJ1 Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          FOXJ1 Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          FOXJ1 Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          FOXJ1 Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          FOXJ1 Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          FOXJ1 Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          FOXJ1 Antibody in Immunocytochemistry (ICC/IF)
          FOXJ1 Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          FOXJ1 Antibody in Immunocytochemistry (ICC/IF)
          FOXJ1 Antibody in Immunocytochemistry, Flow Cytometry (ICC/IF, Flow)
          FOXJ1 Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          FOXJ1 Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          FOXJ1 Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          FOXJ1 Antibody in Immunocytochemistry (ICC/IF)
          FOXJ1 Antibody in Immunocytochemistry, Flow Cytometry (ICC/IF, Flow)
          FOXJ1 Antibody in Immunohistochemistry (Frozen), Immunocytochemistry (IHC (F), ICC/IF)
          FOXJ1 Antibody in Immunocytochemistry (ICC/IF)
          FOXJ1 Antibody in Immunocytochemistry (ICC/IF)
          FOXJ1 Antibody in Immunohistochemistry (Frozen), Immunocytochemistry (IHC (F), ICC/IF)
          FOXJ1 Antibody in Immunocytochemistry (ICC/IF)
          FOXJ1 Antibody in Immunocytochemistry (ICC/IF)
          FOXJ1 Antibody in Immunocytochemistry (ICC/IF)
          FOXJ1 Antibody in Immunocytochemistry (ICC/IF)
          FOXJ1 Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          FOXJ1 Antibody in Immunohistochemistry, Immunohistochemistry (PFA fixed), Immunocytochemistry (IHC, IHC (PFA), ICC/IF)
          FOXJ1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FOXJ1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          FOXJ1 Antibody in Immunohistochemistry, Immunohistochemistry (Frozen) (IHC, IHC (F))
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (IHC)
          FOXJ1 Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          FOXJ1 Antibody in Western Blot (WB)
          FOXJ1 Antibody in Western Blot, ChIP Assay (WB, ChIP)
          FOXJ1 Antibody in Western Blot (WB)
          FOXJ1 Antibody in Western Blot (WB)
          FOXJ1 Antibody in Western Blot (WB)
          FOXJ1 Antibody
          Product Details
          14-9965-80
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1-10 µg/mL
          View 6 publications 6 publications
          Immunohistochemistry (IHC)
          -
          View 64 publications 64 publications
          Immunohistochemistry (Paraffin) (IHC (P))
          1-10 µg/mL
          View 6 publications 6 publications
          Immunohistochemistry (PFA fixed) (IHC (PFA))
          -
          View 3 publications 3 publications
          Immunohistochemistry (Frozen) (IHC (F))
          -
          View 5 publications 5 publications
          Immunocytochemistry (ICC/IF)
          Assay-Dependent
          View 35 publications 35 publications
          Flow Cytometry (Flow)
          -
          View 2 publications 2 publications
          ChIP assay (ChIP)
          -
          View 2 publications 2 publications
          Product Specifications
          Species Reactivity
          Human, Mouse
          Published species
          Fruit fly, Human, Mouse, Pig, Rat, Zebrafish
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          2A5
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          0.5 mg/mL
          Amount
          25 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.2
          Contains
          0.09% sodium azide
          Storage conditions
          4°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_1548836
          Product Specific Information
          Description: This 2A5 monoclonal antibody reacts with Foxj1, a 50-kDa member of the Forkhead-box (FOX) family of winged-helix transcription factors. Expression of Foxj1 is high in ciliated epithelial cells and low in naive T and B cells, where cellular activation results in downregulation of the factor. Foxj1 modulates germinal center formation and Th1 activation by repressing NFkappaB through IkappaBbeta induction. As such, this transcription factor has been suggested to antagonize autoimmune reactions such as systemic lupus erythematosus and rheumatoid arthritis. In addition, Foxj1 has been linked to the formation of motile cilia in epithelial cells.
          Applications Reported: This 2A5 antibody has been reported for use in western blotting, immunohistochemical staining of formalin-fixed paraffin embedded tissue sections, and immunocytochemistry.
          Applications Tested: This 2A5 antibody has been tested by western blot of mouse tracheal epithelial cells and testes, as well as by immunohistochemistry of mouse lung tissue. This antibody can be used between 1-10 µg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.
          Purity: Greater than 90%, as determined by SDS-PAGE.
          Aggregation: Less than 10%, as determined by HPLC.
          Filtration: 0.2 µm post-manufacturing filtered.
          Target Information
          Forkhead-box J1 (FOXJ1) is a 421-amino acid transcription factor that suppresses T cell activity and spontaneous autoimmunity, through the repression of NF B activity. FOXJ1 also inhibits the humoral immune response in B cells. FOXJ1 deficiency in B cells results in spontaneous and accentuated germinal center formation and is implicated in the development of pathogenic autoantibodies and accentuated responses to immunizations. Abnormal expression of FOXJ1 may be associated with autoimmune diseases and/or other inflammatory diseases. FOXJ1 is also required for cilia formation and left-right axis determination because it increases calpastatin expression, a protein necessary for the ability of basal bodies to anchor to the apical cytoskeleton. FOXJ1 expression may function as an early marker of epithelial cell differentiation, recovery, and function.
          Bioinformatics
          Protein Aliases: fork head homologue 4; Forkhead box protein J1; forkhead transcription factor HFH-4; forkhead-like 13; Forkhead-related protein FKHL13; Hepatocyte nuclear factor 3 forkhead homolog 4; HFH-4; HNF-3/forkhead homolog 4; MGC35202
          View more View less
          Gene Aliases: CILD43; FKHL-13; FKHL13; FOXJ1; HFH-4; HFH4
          View more View less
          UniProt ID: (Human) Q92949, (Mouse) Q61660
          View more View less
          Entrez Gene ID: (Human) 2302, (Mouse) 15223
          View more View less
          Function(s)
          transcription regulatory region sequence-specific DNA binding RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding DNA binding transcription factor activity, sequence-specific DNA binding protein binding sequence-specific DNA binding transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter central tolerance induction negative regulation of germinal center formation positive regulation of central B cell tolerance induction negative regulation of humoral immune response mediated by circulating immunoglobulin regulation of transcription, DNA-templated regulation of transcription from RNA polymerase II promoter humoral immune response spermatogenesis determination of left/right symmetry pattern specification process brain development heart development protein localization cell projection organization actin cytoskeleton organization regulation of epithelial cell differentiation ciliary basal body organization negative regulation of interleukin-6 production negative regulation of T cell differentiation in thymus axoneme assembly establishment of apical/basal cell polarity metanephric part of ureteric bud development negative regulation of T cell proliferation motile cilium assembly positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter cell maturation negative regulation of B cell activation leukocyte migration cilium morphogenesis lung epithelium development epithelium development left/right pattern formation glomerular parietal epithelial cell development negative regulation of NIK/NF-kappaB signaling positive regulation of lung ciliated cell differentiation transcription, DNA-templated transcription from RNA polymerase II promoter cell differentiation negative regulation of NF-kappaB transcription factor activity cilium assembly negative regulation of interleukin-6 biosynthetic process activation of GTPase activity
          Spain flag icon
          Spain
          TEST
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline