    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • GLIS3 Antibodies
          NeoBiotechnologies
          GLIS family zinc finger 3 (GLIS3)/ZNF515 Monoclonal Antibody (PCRP-GLIS3-1B11)
          View all (7) GLIS3 antibodies
          Tech Support
          Cite GLIS family zinc finger 3 (GLIS3)/ZNF515 Monoclonal Antibody (PCRP-GLIS3-1B11)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (3)
          GLIS family zinc finger 3 (GLIS3)/ZNF515 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          GLIS family zinc finger 3 (GLIS3)/ZNF515 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          FIGURE: 1 / 3
          GLIS family zinc finger 3 (GLIS3)/ZNF515 Antibody (169792-MSM1-P1) in Flow
          Flow cytometric analysis of PFA-fixed HeLa cells. GLIS3 Mouse Monoclonal Antibody (PCRP-GLIS3-1B11) followed by goat anti-mouse IgG-CF488 (blue); isotype control (red). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          GLIS family zinc finger 3 (GLIS3)/ZNF515 Antibody in Flow Cytometry (Flow)
          GLIS family zinc finger 3 (GLIS3)/ZNF515 Antibody in Peptide array (ARRAY)
          GLIS family zinc finger 3 (GLIS3)/ZNF515 Antibody in SDS-PAGE (SDS-PAGE)
          GLIS family zinc finger 3 (GLIS3)/ZNF515 Monoclonal Antibody (PCRP-GLIS3-1B11)
          Product Details
          169792-MSM1-P1
          Applications
          Tested Dilution
          Publications
          Flow Cytometry (Flow)
          1-2 µg/10^6 cells
          -
          ELISA (ELISA)
          Assay-dependent
          -
          Immunoprecipitation (IP)
          1-2 g/500 µg lysate
          -
          Peptide Array (Array)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG1, kappa
          Class
          Monoclonal
          Type
          Antibody
          Clone
          PCRP-GLIS3-1B11
          Immunogen
          Recombinant full-length human GLIS3 protein
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          200 µg/mL
          Amount
          100 µg
          Purification
          Protein A/G
          Storage buffer
          PBS, pH 7.4, with 0.05% BSA
          Contains
          0.05% sodium azide
          Storage conditions
          4°C, do not freeze
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          This gene is a member of the GLI-similar zinc finger protein family and encodes a nuclear protein with five C2H2-type zinc finger domains. This protein functions as both a repressor and activator of transcription and is specifically involved in the development of pancreatic beta cells, the thyroid, eye, liver and kidney. Mutations in this gene have been associated with neonatal diabetes and congenital hypothyroidism (NDH). Alternatively spliced variants that encode different protein isoforms have been described but the full-length nature of only two have been determined.
          Bioinformatics
          Protein Aliases: GLI-similar 3; OK/KNS-cl.4; Zinc finger protein 515; Zinc finger protein GLIS3
          View more View less
          Gene Aliases: GLIS3; NDH; ZNF515
          View more View less
          UniProt ID: (Human) Q8NEA6
          View more View less
          Entrez Gene ID: (Human) 169792
          View more View less
          Function(s)
          RNA polymerase II regulatory region sequence-specific DNA binding RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding DNA binding protein binding zinc ion binding metal ion binding sequence-specific double-stranded DNA binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter regulation of transcription from RNA polymerase II promoter transcription from RNA polymerase II promoter positive regulation of transcription from RNA polymerase II promoter
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Privacy Information Center
          • Legal Notice
          Germany flag icon
          Germany
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline